• ELISA Kits ›
  • Caspase 8 ELISA Kits

Invitrogen

Human Caspase 8/FLICE ELISA Kit

1 Reference

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Technical guide & protocol Tech Support
Technical guide & protocol Tech Support
Technical guide & protocol Tech Support
Human Caspase 8/FLICE ELISA Kit
Group 53 Created with Sketch.
Human Caspase 8/FLICE ELISA Kit
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Human Caspase 8/FLICE ELISA Kit

Representative standard curve for Human Caspase-8 ELISA {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Human Caspase 8/FLICE ELISA Kit
Human Caspase 8/FLICE ELISA Kit (BMS2024)
videoThumbnail
►

Product Specifications

Analytical sensitivity

0.10 ng/mL

Assay range

0.16-10.0 ng/mL

Sample type/volume

Cell Lysate
50 µL
Serum
50 µL
Supernatant
50 µL

Hands-on time

1 hr 20 min

Time-to-result

3 hr 15 min

Homogenous (no wash)

No

Interassay CV

8.5%

Intraassay CV

6.7%

Instrument

Absorbance Microplate Reader

Product size

96 Tests

Contents

Pre-coated 96 well plate
Standard
Sample Diluent
Assay Buffer concentrate
Detection Antibody
anti-rabbit HRP conjugate
Lysis Buffer
Wash Buffer
Chromogen
Stop Solution
Adhesive Plate Covers

Shipping conditions

Wet ice

Storage

2-8°C

Protein name

Caspase 8

Species (tested)

Human

Assay kit format

Sandwich ELISA Kit

Detector antibody conjugate

HRP

Label or dye

HRP

About This Kit

The Human Caspase 8/FLICE ELISA quantitates Hu Caspase 8 in human serum, cell lysate or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Caspase 8.

Principle of the method

The Human Caspase 8 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Rigorous validation

Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Target information

Caspase 8 (CASP8) is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases play a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes composed of a pro-domain, a large protease subunit, and a small protease subunit. Activation of caspases requires proteolytic processing at conserved internal aspartic residues to generate a heterodimeric enzyme consisting of the large and small subunits. Caspase 8 is involved in the programmed cell death induced by Fas and various apoptotic stimuli. The N-terminal FADD-like death effector domain of Caspase 8 suggests that it may interact with Fas-interacting protein FADD. Caspase 8 was detected in the insoluble fraction of the affected brain region from Huntington disease patients but not in those from normal controls, which implicated the role in neurodegenerative diseases. Caspase 8 binds to the death effector domain (DED) of FADD through an analogous DED domain present in tandem in the pro-form of the Caspase 8 protein. Activated Caspase 8 then activates other downstream caspases including Caspase 9, thereby committing the cell to undergo apoptosis. In addition, Caspase 8 also reacts with Jurkat cells and Tonsil. Overexpression of Caspase 8 induces apoptosis, which can be blocked by inhibitors specific for the ICE family. Many alternatively spliced transcript variants encoding different isoforms have been described for Caspase 8, however, not all variants have had their full-length sequences determined.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Gene aliases : ALPS2B, CAP4, Casp-8, CASP8, FLICE, MACH, MCH5

Gene ID : (Human) 841

Gene symbol : CASP8

Protein Aliases : Apoptotic cysteine protease, Apoptotic protease Mch-5, CAP4, CASP-8, caspase 8, apoptosis-related cysteine peptidase, apoptosis-related cysteine protease, Caspase-8, Caspase8, Caspase-8 precursor, EC 3.4.22.61, FLJ17672, ICE8, MGC78473, OTTHUMP00000163720, OTTHUMP00000165063, OTTHUMP00000206557, OTTHUMP00000206581, FADD-homologous ICE/CED-3-like protease, FADD-like ICE, FLICE, ICE-like apoptotic protease 5, MACH, MACH-alpha-1/2/3 protein, MACH-beta-1/2/3/4 protein, MORT1-associated ced-3 homolog

UniProt ID (Human) Q14790

Recommended Products

IL-6 Human Matched Antibody Pair

Because you are interested in IL-6 Human ELISA Kit.

SIZE

10 x 96 tests

PRICE

USD 400.00

Cat # CHC1263

guarantee_icon

Performance Guarantee

If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0