• ELISA Kits ›
  • CRELD2 ELISA Kits

Invitrogen

Human CRELD2 ELISA Kit

View all (2) CRELD2 ELISA Kits

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Technical guide & protocol Tech Support
Technical guide & protocol Tech Support
Technical guide & protocol Tech Support
Human CRELD2 ELISA Kit
Group 53 Created with Sketch.
Human CRELD2 ELISA Kit
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Human CRELD2 ELISA Kit

ELISA Standard curve of CRELD2 using Human CRELD2 ELISA Kit (Product # EH128RB). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Human CRELD2 ELISA Kit
Human CRELD2 ELISA Kit (EH128RB)
videoThumbnail
►

Product Specifications

Analytical sensitivity

0.13 ng/mL

Assay range

0.137-100 ng/mL

Sample type/volume

Plasma
50 µL
Serum
50 µL
Supernatant
50 µL

Hands-on time

1 hr 20 min

Time-to-result

4 hr 45 min

Homogenous (no wash)

No

Interassay CV

<12%

Intraassay CV

<10%

Instrument

Absorbance Microplate Reader

Product size

96 Tests

Contents

Pre-coated 96 well plate
Standard
Assay Diluent concentrate
Biotinylated Detection Antibody
SAV-HRP
Wash Buffer
Chromogen
Stop Solution
Adhesive Plate Covers

Shipping conditions

Wet ice

Storage

Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C

Protein name

CRELD2

Species (tested)

Human

Assay kit format

Sandwich ELISA Kit

Detector antibody conjugate

Biotin

Label or dye

HRP

About This Kit

Human CRELD2 quantitates human CRELD2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CRELD2.

Target information

CRELD2 may regulate transport of alpha4-beta2 neuronal acetylcholine receptor.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Gene aliases : CRELD2, UNQ185/PRO211

Gene ID : (Human) 79174

Gene symbol : CRELD2

Protein Aliases : CREL2, DKFZp667O055, MGC11256, cysteine rich with EGF-like domains 2, Cysteine-rich with EGF-like domain protein 2, cysteine-rich with EGF-like domains 2, Protein disulfide isomerase CRELD2

UniProt ID (Human) Q6UXH1

Recommended Products

IL-6 Human Matched Antibody Pair

Because you are interested in IL-6 Human ELISA Kit.

SIZE

10 x 96 tests

PRICE

USD 400.00

Cat # CHC1263

guarantee_icon

Performance Guarantee

If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0