• ELISA Kits ›
  • CD300e ELISA Kits

Invitrogen

Human CD300e/LMIR6 ELISA Kit

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Technical guide & protocol Tech Support
Technical guide & protocol Tech Support
Technical guide & protocol Tech Support
Human CD300e/LMIR6 ELISA Kit
Group 53 Created with Sketch.
Human CD300e/LMIR6 ELISA Kit
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Human CD300e/LMIR6 ELISA Kit

ELISA Standard curve of CD300e/LMIR6 using Human CD300e/LMIR6 ELISA Kit (Product # EH85RB). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Human CD300e/LMIR6 ELISA Kit
Human CD300e/LMIR6 ELISA Kit (EH85RB)
videoThumbnail
►

Product Specifications

Analytical sensitivity

0.5 ng/mL

Assay range

0.5-30 ng/mL

Sample type/volume

Serum
50 µL
Plasma
50 µL
Supernatant
50 µL

Hands-on time

1 hr 20 min

Time-to-result

4 hr 45 min

Homogenous (no wash)

No

Interassay CV

<12%

Intraassay CV

<10%

Instrument

Absorbance Microplate Reader

Product size

96 Tests

Contents

Pre-coated 96 well plate
Standard
Assay Diluent concentrate
Biotinylated Detection Antibody
SAV-HRP
Wash Buffer
Chromogen
Stop Solution
Adhesive Plate Covers

Shipping conditions

Wet ice

Storage

Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C

Protein name

CD300e

Species (tested)

Human

Assay kit format

Sandwich ELISA Kit

Detector antibody conjugate

Biotin

Label or dye

HRP

About This Kit

Human CD300e/LMIR6 quantitates human CD300e/LMIR6 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CD300e/LMIR6.

Target information

CD300e is a member of the glycoprotein family of cell surface proteins expressed on myeloid cells. CD300e interacts with the TYRO protein tyrosine kinase-binding protein and is thought to be involved as an activating receptor. CD300e is expressed on mature myeloid dendritic cells and monocytes, and is down regulated on immature dendritic cells. Further, CD300e interacts with DAP-12, which mediates activating signals. The alloreactive response of naïve T cells has been reported to become enhanced by the activation of CD300e. Diseases associated with CD300e dysfunction include tungiasis and lice infestation.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Gene aliases : CD300E, CD300LE, CLM-2, CLM2, CMRF35-A5, CMRF35A5, IREM-2, IREM2, PIgR-2, PIgR2

Gene ID : (Human) 342510

Gene symbol : CD300E

Protein Aliases : CD300 antigen like family member E, CD300 antigen-like family member E, CD300e, CD300e antigen, CLM-2, CMRF35-A5, CMRF35-like molecule 2, Immune receptor expressed on myeloid cells 2, IREM-2, PIgR-2, poly-Ig receptor 2, Polymeric immunoglobulin receptor 2

UniProt ID (Human) Q496F6

Recommended Products

IL-6 Human Matched Antibody Pair

Because you are interested in IL-6 Human ELISA Kit.

SIZE

10 x 96 tests

PRICE

USD 400.00

Cat # CHC1263

guarantee_icon

Performance Guarantee

If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0