• ELISA Kits ›
  • CD147 ELISA Kits

Invitrogen

Human CD147/EMMPRIN ELISA Kit

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Technical guide & protocol Tech Support
Technical guide & protocol Tech Support
Technical guide & protocol Tech Support
Human CD147/EMMPRIN ELISA Kit
Group 53 Created with Sketch.
Human CD147/EMMPRIN ELISA Kit
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Human CD147/EMMPRIN ELISA Kit

ELISA Standard curve of CD147/EMMPRIN using Human CD147/EMMPRIN ELISA Kit (Product # EH78RB). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Human CD147/EMMPRIN ELISA Kit
Human CD147/EMMPRIN ELISA Kit (EH78RB)
videoThumbnail
►

Product Specifications

Analytical sensitivity

12 pg/mL

Assay range

12-2,500 pg/mL

Sample type/volume

Serum
50 µL
Plasma
50 µL
Supernatant
50 µL

Hands-on time

1 hr 20 min

Time-to-result

4 hr 45 min

Homogenous (no wash)

No

Interassay CV

<12%

Intraassay CV

<10%

Instrument

Absorbance Microplate Reader

Product size

96 Tests

Contents

Pre-coated 96 well plate
Standard
Assay Diluent concentrate
Biotinylated Detection Antibody
SAV-HRP
Wash Buffer
Chromogen
Stop Solution
Adhesive Plate Covers

Shipping conditions

Wet ice

Storage

Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C

Protein name

CD147

Species (tested)

Human

Assay kit format

Sandwich ELISA Kit

Detector antibody conjugate

Biotin

Label or dye

HRP

About This Kit

Human CD147/EMMPRIN quantitates human CD147/EMMPRIN in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CD147/EMMPRIN.

Target information

CD147 (basigin, neurothelin, OX-47, 5A11, CE9, M6, EMMPRIN, extracellular matrix metalloproteinase inducer, TCSF, tumour cell-derived collagenase-stimulatory factor) is a 50-60 kDa glycoprotein with multiple glycosylated forms. CD147 is a plasma membrane protein that is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. CD147 is also a member of the immunoglobulin superfamily. The highest level of CD147 expression is on metabolically active cells, such as lymphoblasts, inflammatory cells, brown adipocytes and malignant tumor cells. CD147 has multiple functions, including facilitating of cell surface expression of monocarboxylate transporter proteins and extracellular matrix metalloproteinases, regulation of integrin functions, it plays roles in cell development and activation, fetal development or retinal function.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Gene aliases : 5F7, BSG, CD147, EMMPRIN, OK, TCSF, UNQ6505/PRO21383

Gene ID : (Human) 682

Gene symbol : BSG

Protein Aliases : 5F7, Basigin, CD147, Collagenase stimulatory factor, EMMPRIN, Extracellular matrix metalloproteinase inducer, HAb18G, Hepatoma-associated antigen, Leukocyte activation antigen M6, OK blood group antigen, TCSF, Tumor cell-derived collagenase stimulatory factor

UniProt ID (Human) P35613

Recommended Products

IL-6 Human Matched Antibody Pair

Because you are interested in IL-6 Human ELISA Kit.

SIZE

10 x 96 tests

PRICE

USD 400.00

Cat # CHC1263

guarantee_icon

Performance Guarantee

If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0