• ELISA Kits ›
  • Aldosterone ELISA Kits
Invitrogen
Aldosterone Competitive ELISA Kit
1 Reference
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Technical guide & protocol Tech Support
Technical guide & protocol Tech Support
Technical guide & protocol Tech Support
Aldosterone Competitive ELISA Kit (EIAALD)
Group 53 Created with Sketch.
Aldosterone Competitive ELISA Kit (EIAALD)
Group 53 Created with Sketch.
FIGURE: 1 / 2
{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}
Aldosterone Competitive ELISA Kit
Competitive ELISA {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product
{{$ctrl.videoDesc}}
{{$ctrl.videoLongDesc}}
Aldosterone Competitive ELISA Kit (EIAALD)
videoThumbnail
►
Product Specifications
Analytical sensitivity
4.97 pg/mL
Assay range
3.906-4,000 pg/mL
Sample type/volume
Fecal Extract
100 µL
Plasma
100 µL
Serum
100 µL
Supernatant
100 µL
Urine
25 µL
Hands-on time
1 hr 20 min
Time-to-result
16 hr 45 min
Homogenous (no wash)
No
Interassay CV
19.5%
Intraassay CV
6.9%
Instrument
Absorbance Microplate Reader
Product size
96 Tests
Contents
Pre-coated 96 well plate
Standard
Assay Diluent concentrate
HRP-Conjugated Detection Antibody
Wash Buffer
Chromogen
Stop Solution
Adhesive Plate Covers
Shipping conditions
Wet ice
Storage
-20°C
Protein name
Aldosterone
Species (tested)
Chemical
Assay kit format
Competitive ELISA
Detector antibody conjugate
HRP
Label or dye
HRP
About This Kit
The Aldosterone Competitive ELISA quantitates aldosterone in extracted serum, plasma, urine, dried fecal extracts or cell culture medium.
Principle of the method
The Aldosterone Competitive ELISA Kit research-use-only kit is designed to quantitatively measure aldosterone present in extracted serum and plasma, or in urine, extracted dried fecal samples, and tissue culture media samples. An aldosterone stock solution is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into the provided plate coated with a capture antibody. An aldosterone-peroxidase conjugate is added to the standards and samples in the wells. The binding reaction is initiated by the addition of a polyclonal antibody to aldosterone to each well. After incubation, the plate is washed and substrate is added. The substrate reacts with the bound aldosterone-peroxidase conjugate. After a short incubation, the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader capable of measuring at 450 nm.
Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Target information
Aldosterone is a hormone produced by the adrenal glands that helps regulate blood pressure through increasing both the release of potassium and the uptake of water and sodium by the kidneys. It is essential for sodium conservation in the kidney, salivary glands, sweat glands and colon. It plays a central role in the regulation of the plasma sodium (Na+), the extracellular potassium (K+) and arterial blood pressure. It does so mainly by acting on the mineralocorticoid receptors in the distal tubules and collecting ducts of the nephron. It influences the reabsorption of sodium and excretion of potassium (from and into the tubular fluids, respectively) of the kidney, thereby indirectly influencing water retention or loss, blood pressure and blood volume. When dysregulated, aldosterone is pathogenic and contributes to the development and progression of cardiovascular and renal disease. Aldosterone has exactly the opposite function of the atrial natriuretic hormone secreted by the heart.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Bioinformatics
Protein Aliases : C21H28O5
Recommended Products
IL-6 Human Matched Antibody Pair
Because you are interested in IL-6 Human ELISA Kit.
SIZE
10 x 96 tests
PRICE
USD 400.00
Cat # CHC1263
guarantee_icon
Performance Guarantee
If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
help_icon
We're here to help
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support
Your items have has been added!
Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0