Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • c-Kit (CD117) Antibodies

Fabgennix

cKit Polyclonal Antibody, FITC

View all (210) c-Kit (CD117) antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite cKit Polyclonal Antibody, FITC

cKit Polyclonal Antibody, FITC

Product Details

CKIT-112-FITC

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:1,000
-

Immunohistochemistry (IHC)

1:50-1:250
-

ELISA (ELISA)

1:10,000
-

Immunoprecipitation (IP)

1:50-1:250
-
Product Specifications

Species Reactivity

Dog, Human, Non-human primate, Pig

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Synthetic peptide taken within amino acid region 430-480 on human Tyrosine-Kinase c-Kit protein.
View immunogen

Conjugate

FITC FITC FITC

Excitation/Emission Max

498/517 nm View spectra spectra

Form

Liquid

Concentration

0.5-1.5 mg/mL

Purification

Affinity chromatography

Storage buffer

proprietary buffer, pH 7.4-7.8, with 0.5% BSA, 30% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C, store in dark

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

KIT (c-KIT) is a proto-oncogene and a type 3 transmembrane receptor for MGF (mast cell growth factor, also known as stem cell factor). KIT was first identified as the cellular homolog of the feline sarcoma viral oncogene v-kit. KIT together with its ligand regulates growth and activation of a variety of hemopoietic and non-hemopoietic cells. Mutations in KIT are associated with gastrointestinal stromal tumors, mast cell disease, acute myelogenous leukemia, and piebaldism. Recently, deregulation of the KIT receptor TK by the prevalent activation loop mutation D816V has served as a focal point in therapeutic strategies aimed at curbing neoplastic mast cell growth. c-Kit is expressed in hematopoietic stem cells, germ cells, mast cells and gastrointestinal tract cajal cells. Upon binding of its ligand stem cell factor (SCF), c-kit dimerizes, resulting in receptor activation and autophosphorylation of various tyrosine residues including tyrosine 703 located on the cytoplasmic domain of the receptor. This modification allows docking of Grb2 and activation of the Ras/ERK signaling pathway. SCF/c-kit can activate multiple downstream signaling pathways including PI3K, PLC-gamma and JAK/STAT. c-kit receptor activation is essential for hematopoiesis, stem cell maintenance and gametogenesis.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: c-kit; c-KIT gene1; CD117; ckit; Mast/stem cell growth factor receptor Kit; p145 c-kit; PBT; Piebald trait protein; Proto-oncogene c-Kit; proto-oncogene tyrosine-protein kinase Kit; receptor tyrosine kinase c-kit; SCFR; soluble KIT variant 1; Tyrosine-protein kinase Kit; v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog; v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein

View more View less

Gene Aliases: C-Kit; CD117; KIT; MGF; PBT; SCFR

View more View less

UniProt ID: (Dog) O97799, (Human) P10721, (Pig) Q2HWD6

View more View less

Entrez Gene ID: (Dog) 403811, (Human) 3815, (Pig) 396810

View more View less

Function(s)
protease binding transmembrane receptor protein tyrosine kinase activity receptor signaling protein tyrosine kinase activity stem cell factor receptor activity ATP binding cytokine binding protein homodimerization activity metal ion binding protein tyrosine kinase activity Ras guanyl-nucleotide exchange factor activity protein binding phosphatidylinositol-4,5-bisphosphate 3-kinase activity
Process(es)
activation of MAPK activity ovarian follicle development myeloid progenitor cell differentiation lymphoid progenitor cell differentiation immature B cell differentiation dendritic cell cytokine production mast cell chemotaxis glycosphingolipid metabolic process inflammatory response spermatogenesis spermatid development positive regulation of cell proliferation regulation of cell shape male gonad development response to radiation peptidyl-tyrosine phosphorylation cytokine-mediated signaling pathway signal transduction by protein phosphorylation lamellipodium assembly T cell differentiation erythrocyte differentiation melanocyte differentiation actin cytoskeleton reorganization mast cell cytokine production embryonic hemopoiesis ectopic germ cell programmed cell death intracellular signal transduction hematopoietic stem cell migration megakaryocyte development Fc receptor signaling pathway Kit signaling pathway erythropoietin-mediated signaling pathway positive regulation of tyrosine phosphorylation of Stat1 protein positive regulation of tyrosine phosphorylation of Stat3 protein positive regulation of tyrosine phosphorylation of Stat5 protein negative regulation of programmed cell death mast cell degranulation pigmentation protein autophosphorylation regulation of developmental pigmentation digestive tract development stem cell differentiation detection of mechanical stimulus involved in sensory perception of sound positive regulation of sequence-specific DNA binding transcription factor activity cell chemotaxis mast cell differentiation cellular response to thyroid hormone stimulus melanocyte migration melanocyte adhesion MAPK cascade signal transduction germ cell migration visual learning positive regulation of gene expression positive regulation of phospholipase C activity regulation of phosphatidylinositol 3-kinase signaling positive regulation of phosphatidylinositol 3-kinase signaling stem cell population maintenance hemopoiesis positive regulation of cell migration positive regulation of pseudopodium assembly somatic stem cell population maintenance regulation of cell proliferation positive regulation of MAPK cascade positive regulation of GTPase activity positive regulation of phosphatidylinositol 3-kinase activity positive regulation of Notch signaling pathway positive regulation of JAK-STAT cascade phosphatidylinositol phosphorylation phosphatidylinositol-mediated signaling somatic stem cell division positive regulation of long-term neuronal synaptic plasticity epithelial cell proliferation mast cell proliferation positive regulation of vascular smooth muscle cell differentiation
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0