Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • alpha Synuclein Antibodies

Invitrogen

alpha Synuclein Monoclonal Antibody (10H7), PerCP

View all (83) alpha Synuclein antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite alpha Synuclein Monoclonal Antibody (10H7), PerCP

  • Antibody Testing Data (2)
alpha Synuclein Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
alpha Synuclein Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

alpha Synuclein Antibody (MA5-45842) in ICC/IF

Immunocytochemistry/Immunofluorescence analysis using human neuroblastoma cells. Fixation involved 4% Formaldehyde for 15 min at RT. Samples were incubated with Alpha Synuclein monoclonal antibody (Product # MA5-45842) at 1:100 for 60 min at RT, followed by Goat Anti-Mouse ATTO 488 at 1:200 for 60 min at RT. Counterstain used was Phalloidin Texas Red F-Actin stain; DAPI (blue) nuclear stain at 1:1,000, 1:5,... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

alpha Synuclein Antibody in Immunocytochemistry (ICC/IF)
alpha Synuclein Antibody in Western Blot (WB)
alpha Synuclein Monoclonal Antibody (10H7), PerCP

Product Details

MA5-45842

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000
-

Immunocytochemistry (ICC/IF)

1:100
-

ELISA (ELISA)

1:1,000
-

Dot blot (DB)

1:1,000
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

10H7

Immunogen

Mouse alpha synuclein aggregate
View immunogen

Conjugate

PerCP PerCP PerCP
  • APC 
  • FITC
  • Request custom conjugation

Excitation/Emission Max

482/675 nm View spectra spectra

Form

Liquid

Concentration

1 mg/mL

Purification

Protein G

Storage buffer

95.64mM phosphate/2.48mM MES, pH 7.4, with 0.5M EDTA

Contains

no preservative

Storage conditions

4°C

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2932296

Product Specific Information

A 1:1,000 dilution of MA5-45842 was sufficient for detection of Alpha Synuclein in 15 µg of human brain cell lysate by ECL immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody.|Binds within the 101-130 aa region of human alpha synuclein, determined using Dot blot and ELISA with corresponding peptides. Detects approximately 14 kDa band corresponding to Alpha synuclein. Band at approximately 40 kDa is a trimer. Does not detect truncated alpha Synuclein (aa 1-114)

Target Information

Alpha-synuclein is a member of the synuclein family, which also includes beta- and gamma-synuclein. These proteins are abundant in the brain and play a role in regulating synaptic transmission and membrane trafficking. Alpha- and beta-synuclein have been shown to selectively inhibit phospholipase D2, suggesting that they may also be involved in signaling pathways. Defects in the SNCA gene, which encodes alpha-synuclein, have been implicated in the pathogenesis of Parkinson's disease. Additionally, alpha-synuclein peptides are a major component of amyloid plaques in the brains of patients with Alzheimer's disease. Multiple alternatively spliced transcripts encoding different isoforms have been identified for this gene. Diseases associated with SNCA include Parkinson Disease 1, Autosomal Dominant and Dementia, Lewy Body.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Alpha synuclein; Alpha-synuclein; NACP; non A-beta component of AD amyloid; Non-A beta component of AD amyloid; non-A4 component of amyloid; Non-A4 component of amyloid precursor; synuclein alpha-140; synuclein, alpha (non A4 component of amyloid precursor)

View more View less

Gene Aliases: alphaSYN; NACP; PARK1; PARK4; PD1; SNCA; Syn

View more View less

UniProt ID: (Human) P37840, (Rat) P37377, (Mouse) O55042

View more View less

Entrez Gene ID: (Human) 6622, (Rat) 29219, (Mouse) 20617

View more View less

Function(s)
magnesium ion binding fatty acid binding copper ion binding calcium ion binding protein binding phospholipid binding microtubule binding ferrous iron binding zinc ion binding oxidoreductase activity kinesin binding protein domain specific binding histone binding identical protein binding alpha-tubulin binding cysteine-type endopeptidase inhibitor activity involved in apoptotic process phospholipase binding transcription regulatory region DNA binding dynein binding protein N-terminus binding tau protein binding beta-tubulin binding phosphoprotein binding phospholipase D inhibitor activity enzyme binding Hsp70 protein binding arachidonic acid binding metal ion binding
Process(es)
negative regulation of transcription from RNA polymerase II promoter microglial cell activation positive regulation of receptor recycling negative regulation of protein phosphorylation positive regulation of neurotransmitter secretion fatty acid metabolic process neutral lipid metabolic process phospholipid metabolic process apoptotic process activation of cysteine-type endopeptidase activity involved in apoptotic process mitochondrial membrane organization aging adult locomotory behavior response to iron(II) ion regulation of phospholipase activity negative regulation of platelet-derived growth factor receptor signaling pathway regulation of glutamate secretion regulation of dopamine secretion negative regulation of microtubule polymerization receptor internalization protein destabilization response to magnesium ion negative regulation of transporter activity response to lipopolysaccharide negative regulation of monooxygenase activity positive regulation of peptidyl-serine phosphorylation response to interferon-gamma cellular response to oxidative stress negative regulation of histone acetylation regulation of locomotion dopamine biosynthetic process response to drug mitochondrial ATP synthesis coupled electron transport regulation of macrophage activation positive regulation of apoptotic process negative regulation of apoptotic process negative regulation of cysteine-type endopeptidase activity involved in apoptotic process extracellular fibril organization negative regulation of neuron apoptotic process cellular protein metabolic process cellular response to fibroblast growth factor stimulus positive regulation of endocytosis negative regulation of exocytosis negative regulation of dopamine metabolic process behavioral response to cocaine regulation of long-term neuronal synaptic plasticity synaptic vesicle endocytosis synapse organization regulation of acyl-CoA biosynthetic process positive regulation of release of sequestered calcium ion into cytosol dopamine uptake involved in synaptic transmission negative regulation of dopamine uptake involved in synaptic transmission negative regulation of serotonin uptake negative regulation of norepinephrine uptake calcium ion homeostasis oxidation-reduction process excitatory postsynaptic potential long-term synaptic potentiation positive regulation of inositol phosphate biosynthetic process negative regulation of thrombin receptor signaling pathway response to interleukin-1 cellular response to copper ion cellular response to epinephrine stimulus positive regulation of protein serine/threonine kinase activity negative regulation of neuron death negative regulation of mitochondrial electron transport, NADH to ubiquinone positive regulation of glutathione peroxidase activity positive regulation of hydrogen peroxide catabolic process regulation of synaptic vesicle recycling regulation of reactive oxygen species biosynthetic process negative regulation of chaperone-mediated autophagy synaptic transmission, dopaminergic chemical synaptic transmission response to cocaine dopamine metabolic process regulation of neuron apoptotic process regulation of neurotransmitter secretion regulation of neuronal synaptic plasticity synaptic vesicle transport positive regulation of synaptic transmission membrane organization regulation of neuron death regulation of reactive oxygen species metabolic process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0