Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • alpha-1a Adrenergic Receptor Antibodies

Alomone Labs, LTD

alpha 1A-Adrenergic Receptor (extracellular) Polyclonal Antibody

View all (11) alpha-1a Adrenergic Receptor antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite alpha 1A-Adrenergic Receptor (extracellular) Polyclonal Antibody

  • Antibody Testing Data (3)
alpha 1A-Adrenergic Receptor (extracellular) Antibody in Immunohistochemistry (IHC)
Group 53 Created with Sketch.
alpha 1A-Adrenergic Receptor (extracellular) Antibody in Immunohistochemistry (IHC)
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

alpha 1A-Adrenergic Receptor (extracellular) Antibody (AAR-015-200UL) in IHC

Expression of alpha 1A-Adrenergic receptor in rat cerebellum - Immunohistochemical staining of rat cerebellum using Anti- alpha 1A-Adrenergic Receptor (extracellular) Antibody (#AAR-015), (1:100). A. alpha 1A-adrenoceptor (green) appears in fibers of Bergmann glia (triangle points at an example). B. S100 beta (red), a marker of Bergmann glia, is stained in the same section. C. Merge of the images demonstrat... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

alpha 1A-Adrenergic Receptor (extracellular) Antibody in Immunohistochemistry (IHC)
alpha 1A-Adrenergic Receptor (extracellular) Antibody in Western Blot (WB)
alpha 1A-Adrenergic Receptor (extracellular) Antibody in Flow Cytometry (Flow)
alpha 1A-Adrenergic Receptor (extracellular) Polyclonal Antibody

Product Details

AAR-015-200UL

Applications
Tested Dilution
Publications

Western Blot (WB)

1:200
-

Immunohistochemistry (IHC)

1:100
-

Flow Cytometry (Flow)

2.5 µg
-
Product Specifications

Species Reactivity

Human, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

EDETI*SQINEEPG(C), corresponding to amino acid residues 171-183 of human alpha 1A-adrenoceptor with replacement of cysteine 176 (C176) with serine (*S), 2nd extracellular loop
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

0.7 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.4, with 1% BSA

Contains

0.05% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).

Target Information

Adrenergic receptors (ARs) are members of the 7-transmembrane domain G-protein-coupled receptor superfamily that bind the endogenous catecholamines epinephrine and norepinephrine. Pharmacological, structural, and molecular cloning data indicate significant heterogeneity within this receptor family. Nine receptor subtypes have been identified thus far including three alpha-1 AR subtypes (1A/D, 1B, and 1C), three alpha-2 ARs (2A, 2B, and 2C), and three beta AR subtypes (1, 2, and 3). ARs participate in either the onset or maintenance of several disease states including hypertension, cardiac dysfunction (congestive heart failure, ischemia, arrhythmias), diabetes, glaucoma, depression, and impotence. A1AR subtypes are found in numerous tissues and have been shown to be involved in the regulation of blood pressure due to changes in vascular tone and cardiac output. Effects on uterine contraction, hepatic glucose metabolism, heat shock protein 70 (HSP70), proto-oncogene expression, and mitogenesis have been linked to A1AR activation. Norepinephrine can increase nitric oxide synthase (NOS) levels in the hypothalamus by activating alpha-1 AR which may indirectly suppress the release of luteinizing hormone (LH), the androgens, estrogen, and progesterone.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: ADA1; adrenergic alpha 1c receptor; adrenergic receptor alpha 1c subtype; adrenergic receptor, alpha 1a; adrenergic receptor, alpha 1c; adrenergic, alpha-1A-, receptor; alpha 1A-adrenoceptor; alpha 1A-adrenoreceptor; alpha 1C-adrenergic receptor; Alpha-1A adrenergic receptor; Alpha-1A adrenoceptor; Alpha-1A adrenoreceptor; Alpha-1C adrenergic receptor; Alpha-adrenergic receptor 1c; Alpha1C-AR; G protein coupled receptor

View more View less

Gene Aliases: ADRA1A; ADRA1C; ADRA1L1; ALPHA1AAR

View more View less

UniProt ID: (Human) P35348, (Rat) P43140

View more View less

Entrez Gene ID: (Human) 148, (Rat) 29412

View more View less

Function(s)
alpha1-adrenergic receptor activity protein heterodimerization activity
Process(es)
negative regulation of heart rate involved in baroreceptor response to increased systemic arterial blood pressure norepinephrine-epinephrine vasoconstriction involved in regulation of systemic arterial blood pressure positive regulation of heart rate by epinephrine-norepinephrine positive regulation of the force of heart contraction by epinephrine-norepinephrine positive regulation of systemic arterial blood pressure phospholipase C-activating G-protein coupled receptor signaling pathway activation of phospholipase C activity positive regulation of cytosolic calcium ion concentration adult heart development aging response to hormone positive regulation of heart rate cell growth positive regulation of synaptic transmission, GABAergic negative regulation of Rho protein signal transduction organ growth response to drug positive regulation of MAPK cascade positive regulation of action potential positive regulation of vasoconstriction positive regulation of smooth muscle contraction micturition calcium ion transport into cytosol positive regulation of cardiac muscle contraction positive regulation of ERK1 and ERK2 cascade adrenergic receptor signaling pathway positive regulation of protein kinase C signaling pilomotor reflex apoptotic process smooth muscle contraction signal transduction G-protein coupled receptor signaling pathway cell-cell signaling negative regulation of cell proliferation intracellular signal transduction adenylate cyclase-activating adrenergic receptor signaling pathway
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0