Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: HSLFLRRMR APSKPWSQVD EATIRAIIAV LSTEEQSAGL QQPSGIGQRP RPMSSEELPL ASSWRSNNSR KSSADTEFSD ECTTAERVLM KSPSPALHPP QKYKDRGILH PKRGTEDRSD QSSLKSTDSS SYPSPCASPS PPSSGKGSKS PSPRPNMPVR YFIMKSSNLR NLEISQQKGI WSTTPSNERK LNRAFWESSI VYLVFSVQGS GHFQGFSRMS SEIGREKSQD WGSAGLGGVF KVEWIRKESL PFQFAHHLLN PWNDNKKVQI SRDGQELEPL VGEQLLQLWE RLPLGEKNTT D (1131-1430 aa encoded by BC137285)
Specifically recognizes and binds N6-methyladenosine (m6A)-containing RNAs. M6A is a modification present at internal sites of mRNAs and some non-coding RNAs and plays a role in the efficiency of mRNA splicing, processing and stability.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3'-5' RNA helicase YTHDC2; CsA-associated helicase-like protein; DKFZp564A186; FLJ10053; FLJ2194; hYTHDC2; Keen to exit meiosis leaving testes under-populated protein; Ketu; mYTHDC2; probable ATP-dependent RNA helicase YTHDC2; YTH domain-containing protein 2; YTH domain-containing protein C2
Gene Aliases: 3010002F02Rik; BC037178; CAHL; YTHDC2
UniProt ID: (Human) Q9H6S0, (Mouse) B2RR83
Entrez Gene ID: (Human) 64848, (Mouse) 240255
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support