Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • wit Antibodies

Fabgennix

WIT Wishful Thinking Polyclonal Antibody, FITC

View all (6) wit antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite WIT Wishful Thinking Polyclonal Antibody, FITC

Product Details

WIT-112-FITC

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:1,000
-

Immunohistochemistry (IHC)

1:50-1:250
-

Immunocytochemistry (ICC/IF)

1:50-1:250
-

ELISA (ELISA)

1:10,000
-

Immunoprecipitation (IP)

1:50-1:250
-

Immunomicroscopy (IM)

1:50-1:200
-
Product Specifications

Species Reactivity

Fruit fly

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Synthetic peptide taken within amino acid region 70-120 on WIT Wishful Thinking D. melanogaster protein.

Conjugate

FITC FITC FITC

Excitation/Emission Max

498/517 nm View spectra spectra

Form

Liquid

Concentration

0.5-1.5 mg/mL

Purification

Affinity chromatography

Storage buffer

proprietary buffer, pH 7.4-7.8, with 0.5% BSA, 30% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C, store in dark

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

Wishful thinking (wit) gene, the Drosophila homolog of the vertebrate BMP type II receptor. Mutations in wit result in pharate lethality that can be rescued by expression of a wit transgene in motor neurons but not in muscles. Mutant larvae exhibit small synapses, severe defects in evoked junctional potentials, a lower frequency of spontaneous vesicle release, and an alteration in the ultrastructure of synaptic active zones.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: bone morphogenetic protein; CG10776; CG10776-PA; CG10776-PB; Receptor protein serine/threonine kinase; Serine/threonine kinase-D; wishful thinking; wit; wit-PA; wit-PB

View more View less

Gene Aliases: 1262/15; ALK3/BMPRII; BMP; CG10776; Dmel\CG10776; Dmel_CG10776; l(3)64Aa; l(3)S126215; l(3)SH12; SE20; STK-D; WIT; Wit-C

View more View less

Entrez Gene ID: (Fruit fly) 44096

View more View less

Function(s)
protein kinase activity transmembrane receptor protein serine/threonine kinase activity receptor signaling protein serine/threonine kinase activity transforming growth factor beta receptor activity, type II ATP binding
Process(es)
protein phosphorylation signal transduction transmembrane receptor protein serine/threonine kinase signaling pathway neuromuscular synaptic transmission ventral cord development neuromuscular junction development regulation of synaptic growth at neuromuscular junction positive regulation of synaptic growth at neuromuscular junction lateral inhibition dendrite morphogenesis
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0