Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • VIP Antibodies

OriGene

VIP Monoclonal Antibody (OTI5B5), TrueMAB™

View all (27) VIP antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite VIP Monoclonal Antibody (OTI5B5), TrueMAB™

  • Antibody Testing Data (5)
VIP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
VIP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 5

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

VIP Antibody (CF806852) in IHC (P)

Immunohistochemical staining of paraffin-embedded Carcinoma of Human pancreas tissue using anti-VIP mouse monoclonal antibody. (Heat-induced epitope retrieval by 1mM EDTA in 10mM Tris buffer (pH8.5) at 120°C for 3 min, TA806852)(1:150). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

VIP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
VIP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
VIP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
VIP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
VIP Antibody in Western Blot (WB)
VIP Monoclonal Antibody (OTI5B5), TrueMAB™

Product Details

CF806852

Applications
Tested Dilution
Publications

Western Blot (WB)

1:2,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:150
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

OTI5B5

Immunogen

Full length human recombinant protein of human VIP produced in E.coli.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.3, with 8% trehalose

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)

Target Information

VIP encodes a protein that belongs to the glucagon family. It stimulates myocardial contractility, causes vasodilation, increases glycogenolysis, lowers arterial blood pressure and relaxes the smooth muscle of trachea, stomach and gall bladder. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: MGC13587; PACAP-R-2; prepro-VIP; VIP peptides; VIP-R-1

View more View less

Gene Aliases: PHM27; VIP

View more View less

UniProt ID: (Human) P01282

View more View less

Entrez Gene ID: (Human) 7432

View more View less

Function(s)
hormone activity neuropeptide hormone activity protein binding
Process(es)
positive regulation of endothelial cell proliferation G-protein coupled receptor signaling pathway body fluid secretion learning or memory positive regulation of cell proliferation positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway regulation of protein localization negative regulation of apoptotic process negative regulation of potassium ion transport positive regulation of protein catabolic process positive regulation of vasodilation mRNA stabilization negative regulation of smooth muscle cell proliferation regulation of sensory perception of pain positive regulation of penile erection prolactin secretion
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0