Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • UQCRC1 Antibodies

OriGene

UQCRC1 Monoclonal Antibody (OTI1G2), TrueMAB™

View all (23) UQCRC1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite UQCRC1 Monoclonal Antibody (OTI1G2), TrueMAB™

  • Antibody Testing Data (3)
UQCRC1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
UQCRC1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

UQCRC1 Antibody (TA800033) in ICC/IF

Anti-UQCRC1 mouse monoclonal antibody (TA800033) immunofluorescent staining of COS7 cells transiently transfected by pCMV6-ENTRY UQCRC1(RC203299). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

UQCRC1 Antibody in Immunocytochemistry (ICC/IF)
UQCRC1 Antibody in Western Blot (WB)
UQCRC1 Antibody in Western Blot (WB)
UQCRC1 Monoclonal Antibody (OTI1G2), TrueMAB™

Product Details

TA800033

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:1,000
-

Immunocytochemistry (ICC/IF)

1:100
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

OTI1G2

Immunogen

Human recombinant protein fragment corresponding to amino acids 46-305 of human UQCRC1 produced in E.coli.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS with 1% BSA, 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

UQCRC1 encodes a product that is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. This protein may mediate formation of the complex between cytochromes c and c1.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Complex III subunit 1; Core protein I; Cytochrome b-c1 complex subunit 1, mitochondrial; Ubiquinol-cytochrome-c reductase complex core protein 1

View more View less

Gene Aliases: D3S3191; QCR1; UQCR1; UQCRC1

View more View less

UniProt ID: (Human) P31930

View more View less

Entrez Gene ID: (Human) 7384

View more View less

Function(s)
metalloendopeptidase activity ubiquinol-cytochrome-c reductase activity zinc ion binding ubiquitin protein ligase binding protein complex binding
Process(es)
oxidative phosphorylation mitochondrial electron transport, ubiquinol to cytochrome c aerobic respiration response to activity protein processing response to alkaloid oxidation-reduction process hydrogen ion transmembrane transport
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0