Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • UQCRC1 Antibodies

Proteintech

UQCRC1 Monoclonal Antibody (2G1C11)

1 Reference
View all (23) UQCRC1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite UQCRC1 Monoclonal Antibody (2G1C11)

  • Antibody Testing Data (1)
UQCRC1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
UQCRC1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

UQCRC1 Antibody (67888-1-IG) in WB

Various lysates were subjected to SDS PAGE followed by western blot with 67888-1-Ig (UQCRC1 antibody) at dilution of 1:20000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

UQCRC1 Antibody in Western Blot (WB)

Product Details

67888-1-IG

Applications
Tested Dilution
Publications

Western Blot (WB)

1:20,000
View 1 publication 1 publication
Product Specifications

Species Reactivity

Human, Mouse, Pig, Rat

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

2G1C11

Immunogen

UQCRC1 Fusion Protein Ag17506

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1000 µg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice

Product Specific Information

Aliquoting is unnecessary for -20°C storage.

Target Information

UQCRC1 encodes a product that is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. This protein may mediate formation of the complex between cytochromes c and c1.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: Complex III subunit 1; Core protein I; Cytochrome b-c1 complex subunit 1, mitochondrial; ubiquinol-cytochrome c reductase core protein 1; Ubiquinol-cytochrome-c reductase complex core protein 1

View more View less

Gene Aliases: 1110032G10Rik; D3S3191; QCR1; UQCR1; UQCRC1

View more View less

UniProt ID: (Human) P31930, (Rat) Q68FY0, (Mouse) Q9CZ13

View more View less

Entrez Gene ID: (Human) 7384, (Rat) 301011, (Mouse) 22273

View more View less

Function(s)
metalloendopeptidase activity ubiquinol-cytochrome-c reductase activity zinc ion binding ubiquitin protein ligase binding protein complex binding catalytic activity metal ion binding protein binding
Process(es)
oxidative phosphorylation mitochondrial electron transport, ubiquinol to cytochrome c aerobic respiration response to activity protein processing response to alkaloid oxidation-reduction process hydrogen ion transmembrane transport transport
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0