Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • UAP1 Antibodies

Proteintech

UAP1 Monoclonal Antibody (2C7B2)

3 References
View all (21) UAP1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite UAP1 Monoclonal Antibody (2C7B2)

  • Antibody Testing Data (3)
UAP1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
UAP1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

UAP1 Antibody (67545-1-IG) in ICC/IF

Immunofluorescent analysis of (4% PFA) fixed PC-3 cells using UAP1 antibody (67545-1-Ig, Clone: 2C7B2 ) at dilution of 1:400 and CoraLite®488-Conjugated AffiniPure Goat Anti-Mouse IgG(H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

UAP1 Antibody in Immunocytochemistry (ICC/IF)
UAP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
UAP1 Antibody in Western Blot (WB)

Product Details

67545-1-IG

Applications
Tested Dilution
Publications

Western Blot (WB)

1:10,000
View 2 publications 2 publications

Immunohistochemistry (IHC)

-
View 1 publication 1 publication

Immunohistochemistry (Paraffin) (IHC (P))

1:1,000
-

Immunocytochemistry (ICC/IF)

1:400
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

2C7B2

Immunogen

UAP1 Fusion Protein Ag9513

Conjugate

Unconjugated Unconjugated Unconjugated
  • CoraLite 594
  • CoraLite Plus 488
  • Request custom conjugation

Form

Liquid

Concentration

1000 µg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice

Product Specific Information

Aliquoting is unnecessary for -20°C storage.

Target Information

Converts UDP and GlcNAc-1-P into UDP-GlcNAc, and UDP and GalNAc-1-P into UDP-GalNAc. Isoform AGX1 has 2 to 3 times higher activity towards GalNAc-1-P, while isoform AGX2 has 8 times more activity towards GlcNAc-1-P.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: AGX; Antigen X; Protein-pyrophosphorylation enzyme; sperm associated antigen 2; Sperm-associated antigen 2; testis tissue sperm-binding protein Li 37a; UDP-N-acetylgalactosamine pyrophosphorylase; UDP-N-acetylglucosamine diphosphorylase 1; UDP-N-acetylglucosamine pyrophosphorylase; UDP-N-acetylhexosamine pyrophosphorylase; UDP-N-acetylhexosamine pyrophosphorylase 1

View more View less

Gene Aliases: AGX; AGX1; AGX2; SPAG2; UAP1

View more View less

UniProt ID: (Human) Q16222

View more View less

Entrez Gene ID: (Human) 6675

View more View less

Function(s)
UDP-N-acetylglucosamine diphosphorylase activity carbohydrate binding identical protein binding
Process(es)
UDP-N-acetylglucosamine biosynthetic process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0