Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • TrxR2 Antibodies

Invitrogen

TrxR2 Recombinant Rabbit Monoclonal Antibody (4O1K0)

View all (12) TrxR2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite TrxR2 Recombinant Rabbit Monoclonal Antibody (4O1K0)

  • Antibody Testing Data (1)
TrxR2 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
TrxR2 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

TrxR2 Antibody (MA5-35730) in WB

Western blot analysis of TrxR2 in extracts of various cell lines. Samples were incubated with TrxR2 Monoclonal antibody (Product # MA5-35730) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10,000. Lysates/proteins: 25 µg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Basic Kit. Exposure time: 10s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

TrxR2 Antibody in Western Blot (WB)
TrxR2 Recombinant Rabbit Monoclonal Antibody (4O1K0)

Product Details

MA5-35730

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:2,000
-

ELISA (ELISA)

1 µg/mL
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

4O1K0

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Thioredoxin reductase 2 (TXNRD2 ) (NP_0064312)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.79 mg/mL

Purification

Affinity Chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol, 0.05% BSA

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2849630

Product Specific Information

Immunogen sequence: MAAMAVALRG LGGRFRWRTQ AVAGGVRGAA RGAAAGQRDY DLLVVGGGSG GLACAKEAAQ LGRKVAVVDY VEPSPQGTRW GLGGTCVNVG CIPKKLMHQA

Target Information

The mammalian thioredoxin reductases (TrxRs) are a family of selenocysteine-containing pyridine nucleotide-disulfide oxidoreductases. All the mammalian TrxRs are homologous to glutathione reductase with respect to primary structure including the conserved redox catalytic site (-Cys-Val-Asn-Val-Gly-Cys-) but distinctively with a C-terminal extension containing a catalytically active penultimate selenocysteine (SeCys) residue in the conserved sequence (-Gly-Cys-SeCys-Gly). TrxR is homodimeric protein in which each monomer includes an FAD prosthetic group, a NADPH binding site and a redox catalytic site. Electrons are transferred from NADPH via FAD and the active-site disulfide to C-terminal SeCys-containing redox center, which then reduces the substrate like thioredoxin. The members of TrxR family are 55-58 kilodalton in molecular size and composed of three isoforms including cytosolic TrxR1, mitochondrial TrxR2, and TrxR3, known as Trx and GSSG reductase (TGR). TrxR plays a key role in protection of cells against oxidative stress and redox-regulatory mechanism of transcription factors and various biological phenomena.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Selenoprotein Z; SelZ; Thioredoxin reductase 2, mitochondrial; thioredoxin reductase 3; thioredoxin reductase beta; Thioredoxin reductase TR3; TR beta; TR-beta

View more View less

Gene Aliases: AA118373; ESTM573010; KIAA1652; SELZ; TGR; TR; TR-BETA; TR3; TRXR2; Trxrd2; TXNRD2

View more View less

UniProt ID: (Human) Q9NNW7, (Mouse) Q9JLT4, (Rat) Q9Z0J5

View more View less

Entrez Gene ID: (Human) 10587, (Mouse) 26462, (Rat) 50551

View more View less

Function(s)
thioredoxin-disulfide reductase activity protein binding flavin adenine dinucleotide binding oxidoreductase activity oxidoreductase activity, acting on a sulfur group of donors, NAD(P) as acceptor protein homodimerization activity mercury (II) reductase activity mercury ion binding protein dimerization activity NADP binding
Process(es)
response to oxygen radical heart development hemopoiesis cell redox homeostasis oxidation-reduction process response to selenium ion response to drug response to hydrogen peroxide detoxification of mercury ion response to hyperoxia response to reactive oxygen species cellular oxidant detoxification
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0