Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Transferrin Receptor (CD71) Antibodies

Invitrogen

Transferrin Receptor Polyclonal Antibody

View all (135) Transferrin Receptor (CD71) antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Transferrin Receptor Polyclonal Antibody

  • Antibody Testing Data (4)
Transferrin Receptor Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
Transferrin Receptor Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Transferrin Receptor Antibody (PA5-116065) in IHC (P)

Immunohistochemistry analysis of Transferrin Receptor in rat ovary tissue. Samples were treated with formaldehyde and treated with citrate buffer for antigen retrieval, blocked, and incubated (1.5 hours at 22°C) with polyclonal antibody (Product # PA5-116065) at a dilution of 1:100. Secondary staining was applied with HRP conjugated anti-Rabbit. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Transferrin Receptor Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Transferrin Receptor Antibody in Western Blot (WB)
Transferrin Receptor Antibody in Western Blot (WB)
Transferrin Receptor Antibody in Western Blot (WB)
Transferrin Receptor Polyclonal Antibody

Product Details

PA5-116065

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:2,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:200
-

Immunocytochemistry (ICC/IF)

1:100-1:500
-
Product Specifications

Species Reactivity

Human, Mouse, Non-human primate

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

A synthesized peptide derived from human TFRC(Accession P02786), corresponding to amino acid residues M1-N50.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2900699

Product Specific Information

Antibody detects endogenous levels of total Transferrin Receptor.

Target Information

Cellular uptake of iron occurs via receptor-mediated endocytosis of ligand-occupied transferrin receptor into specialized endosomes. Endosomal acidification leads to iron release. The apotransferrin-receptor complex is then recycled to the cell surface with a return to neutral pH and the concomitant loss of affinity of apotransferrin for its receptor. Transferrin receptor is necessary for development of erythrocytes and the nervous system. Serum transferrin receptor (sTfR) is used as a means of detecting erythropoietin (EPO) misuse by athletes and as a diagnostic test for anemias resulting from a number of conditions including rheumatoid arthritis, pregnancy, irritable bowel syndrome and in HIV patients.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CD antigen CD71; CD71; mammary tumor virus receptor 1; p90; T9; TR; Transferrin receptor protein 1

View more View less

Gene Aliases: 2610028K12Rik; AI195355; AI426448; AU015758; CD71; E430033M20Rik; IMD46; Mtvr-1; Mtvr1; p90; T9; TFR; TFR1; TFRC; TR; TRFR

View more View less

UniProt ID: (Human) P02786, (Mouse) Q62351

View more View less

Entrez Gene ID: (Human) 7037, (Mouse) 22042

View more View less

Function(s)
virus receptor activity glycoprotein binding double-stranded RNA binding transferrin receptor activity protein binding Hsp70 protein binding transferrin transmembrane transporter activity identical protein binding protein homodimerization activity poly(A) RNA binding chaperone binding iron ion transmembrane transporter activity
Process(es)
regulation of cell growth response to hypoxia cellular iron ion homeostasis acute-phase response aging response to nutrient response to iron ion response to manganese ion osteoclast differentiation positive regulation of B cell proliferation receptor internalization response to retinoic acid transferrin transport cellular response to drug positive regulation of T cell proliferation regulation of cell proliferation positive regulation of bone resorption positive regulation of isotype switching response to copper ion viral entry into host cell iron ion import endocytosis receptor-mediated endocytosis iron ion transmembrane transport protein transmembrane transport
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0