Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Torsin 1A Antibodies

Bioss

Torsin A Polyclonal Antibody

View all (9) Torsin 1A antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Torsin A Polyclonal Antibody

  • Antibody Testing Data (1)
Torsin A Antibody in Western Blot (WB)
Group 53 Created with Sketch.
Torsin A Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Torsin A Antibody (BS-11490R) in WB

Lane 1: HL60 cell lysates; Lane 2: MCF-7 cell lysates; Lane 3: K562 cell lysates probed with Torsin A Polyclonal Antibody, Unconjugated (bs-11490R) at 1:1000 dilution and 4°C overnight incubation. Followed by conjugated secondary antibody incubation at 1:20000 for 60 min at 37°C. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Torsin A Antibody in Western Blot (WB)
Torsin A Polyclonal Antibody

Product Details

BS-11490R

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:200
-

Immunohistochemistry (Frozen) (IHC (F))

1:100-1:500
-

Immunocytochemistry (ICC/IF)

1:100-1:500
-

ELISA (ELISA)

1:500-1:1,000
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

KLH conjugated synthetic peptide derived from human Torsin A, amino acids 56-130.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

0.01M TBS, pH 7.4, with 50% glycerol, 1% BSA

Contains

0.02% ProClin 300

Storage conditions

-20°C

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

The protein encoded by this gene is a member of the AAA family of adenosine triphosphatases (ATPases), is related to the Clp protease/heat shock family and is expressed prominently in the substantia nigra pars compacta. Mutations in this gene result in the autosomal dominant disorder, torsion dystonia 1.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: dystonia 1; Dystonia 1 protein; Dystonia 1 torsion; dystonia 1, torsion (autosomal dominant; dystonia 1, torsion (autosomal dominant; torsin A); Torsin 1A; torsin A; Torsin ATPase 1; Torsin ATPase-1A; Torsin family 1 member A; torsin family 1, member A (torsin A); Torsin-1A

View more View less

Gene Aliases: DQ2; DYT1; TA; TOR1A; TORA; torsinA

View more View less

UniProt ID: (Human) O14656, (Rat) Q68G38, (Mouse) Q9ER39

View more View less

Entrez Gene ID: (Human) 1861, (Rat) 266606, (Mouse) 30931

View more View less

Function(s)
protein binding ATP binding cytoskeletal protein binding ATPase activity kinesin binding unfolded protein binding misfolded protein binding nucleotide binding hydrolase activity
Process(es)
protein deneddylation response to oxidative stress organelle organization nuclear envelope organization cell adhesion neuron projection development protein localization to nucleus wound healing, spreading of cells intermediate filament cytoskeleton organization synaptic vesicle transport chaperone mediated protein folding requiring cofactor protein homooligomerization regulation of dopamine uptake involved in synaptic transmission chaperone-mediated protein folding ER-associated misfolded protein catabolic process nuclear membrane organization chaperone-mediated protein transport positive regulation of synaptic vesicle endocytosis regulation of protein localization to cell surface
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0