Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Thyroid Peroxidase Antibodies

Invitrogen

Thyroid Peroxidase Recombinant Rabbit Monoclonal Antibody (313)

View all (49) Thyroid Peroxidase antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Thyroid Peroxidase Recombinant Rabbit Monoclonal Antibody (313)

Thyroid Peroxidase Recombinant Rabbit Monoclonal Antibody (313)

Product Details

MA5-31235

Applications
Tested Dilution
Publications

ELISA (ELISA)

1:5,000-1:10,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

313

Immunogen

Recombinant Human Thyroid peroxidase/TPO Protein (aa 1-846).
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

PBS

Contains

no preservative

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2786878

Product Specific Information

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

This antibody has specificity for Human Thyroid peroxidase/TPO.

Target Information

Thyroid Peroxidase is a gene that encodes for a membrane-bound glycoprotein, which functions as an enzyme and plays a crucial role in the proper functioning of the thyroid gland. This protein is responsible for the iodination of tyrosine residues in thyroglobulin and the formation of phenoxy-ester between pairs of iodinated tyrosines, which leads to the production of thyroid hormones such as thyroxine and triiodothyronine. Mutations in this gene are known to cause several disorders related to thyroid hormone production, including congenital hypothyroidism, congenital goiter, and thyroid hormone organification defect IIA. Several transcript variants encoding distinct isoforms have been identified for this gene, but the full-length nature of some variants remains undetermined. Diseases associated with TPO include Thyroid Dyshormonogenesis 2A and Congenital Hypothyroidism.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: EC 1.11.1; thyroid microsomal antigen; Thyroid peroxidase; thyroperoxidase; TPO; TPXEC 1.11.1.8

View more View less

Gene Aliases: MSA; TDH2A; TPO; TPX

View more View less

UniProt ID: (Human) P07202

View more View less

Entrez Gene ID: (Human) 7173

View more View less

Function(s)
iodide peroxidase activity peroxidase activity calcium ion binding heme binding
Process(es)
thyroid hormone generation response to lipid embryonic hemopoiesis hormone biosynthetic process hydrogen peroxide catabolic process oxidation-reduction process cellular response to nitric oxide cellular oxidant detoxification
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0