Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: APPAPGDFSG EGSQGLPDPS PEPKQLPELI RMKRDGGRLS EADIRGFVAA VVNGSAQGAQ IGAMLMAIRL RGMDLEETSV LTQALAQSGQ QLEWPEAWRQ QLVDKHSTGG VGDKVSLVLA PALAACGCKV PMISGRGLGH TGGTLDKLES IPGFNVIQSP EQMQVLLDQA GCCIVGQSEQ LVPADGILYA ARDVTATVDS LPLITASILS KKLVEGLSAL VVDVKFGGAA VFPNQEQARE LAKTLVGVGA SLGLRVAAAL TAMDKPLGRC VGHALEVEEA LLCMDGAGPP DLRDLVTTLG GALLWLSGHA GTQAQGAARV AAALDDGSAL GRFERMLAAQ GVDPGLARAL CSGSPAERRQ LLPRAREQEE LLAPADGTVE LVRALPLALV LHELGAGRSR AGEPLRLGVG AELLVDVGQR LRRGTPWLRV HRDGPALSGP QSRALQEALV LSDRAPFAAP SPFAELVLPP QQ
Positive Samples: A-549, THP-1
A platelet-derived endothelial growth factor (PD-ECGF), same as thymidine phosphorylase (TP) or gliostatin. In the presence of inorganic orthophosphate, it catalyses the reversible phospholytic cleavage of thymidine and deoxyuridine to their corresponding bases and 2-deoxyribose-1-phosphate. It is both chemotactic and mitogenic for endothelial cells and a non-heparin binding angiogenic factor present in platelets. It is also involved in transformation of fluoropyrimidines, cytotoxic agents used in the treatment of a variety of malignancies, into active cytotoxic metabolites.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gliostatin; OTTHUMP00000196767; OTTHUMP00000196770; PD TP; PD-ECGF; Platelet-derived endothelial cell growth factor; TdRPase; Thymidine phosphorylase; TP
Gene Aliases: ECGF; ECGF1; hPD-ECGF; MEDPS1; MNGIE; MTDPS1; PDECGF; TP; TYMP
UniProt ID: (Human) P19971
Entrez Gene ID: (Human) 1890
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support