Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • TET1 Antibodies

Active Motif

Tet1 Monoclonal Antibody (5D6)

2 References
View all (25) TET1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Tet1 Monoclonal Antibody (5D6)

  • Antibody Testing Data (1)
Tet1 Antibody in ChIP-Sequencing (ChIP-Seq)
Group 53 Created with Sketch.
Tet1 Antibody in ChIP-Sequencing (ChIP-Seq)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Tet1 Antibody (61741) in ChIP-Seq

TET1 antibody tested by ChIP-Seq. ChIP was performed using TET1 antibody with 30 µg chromatin from human brain tumor cells and 4 µl of antibody. ChIP DNA was sequenced on the Illumina HiSeq and 15 million sequence tags were mapped to identify TET1 binding sites. The image shows binding across a region of chromosome 1. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Tet1 Antibody in ChIP-Sequencing (ChIP-Seq)
Tet1 Monoclonal Antibody (5D6)

Product Details

61741

Applications
Tested Dilution
Publications

Western Blot (WB)

-
View 1 publication 1 publication

ChIP-sequencing (ChIP-Seq)

4 µL
-

Miscellaneous PubMed (MISC)

-
View 1 publication 1 publication
Product Specifications

Species Reactivity

Human, Mouse

Published species

Mouse

Host/Isotype

Rat / IgG2a

Class

Monoclonal

Type

Antibody

Clone

5D6

Immunogen

This antibody was raised against a recombinant protein corresponding amino acids 1682-1914 of mouse Tet1.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein G

Storage buffer

PBS, pH 7.5, with 30% glycerol

Contains

0.035% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Wet ice

RRID

AB_2793752

Target Information

Dioxygenase that specifically binds methylcytosine (5mC), a minor base in mammalian DNA found in repetitive DNA elements that is crucial for retrotransposon silencing and mammalian development. Catalyzes the conversion of methylcytosine (5mC) to 5-hydroxymethylcytosine (hmC). The clear function of 5-hydroxymethylcytosine (hmC) is still unclear but it may influence chromatin structure and recruit specific factors or may constitute an intermediate component in cytosine demethylation. 5-hydroxymethylcytosine (hmC) is present in ES cells and is enriched in the brain, especially in Purkinje neurons. May play a role in the fetal development of heart, lung and brain.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: CXXC finger 6; CXXC zinc finger 6; CXXC-type zinc finger protein 6; Leukemia-associated protein with a CXXC domain; Methylcytosine dioxygenase TET1; RP11-119F7.1; Ten-eleven translocation 1 gene protein; Ten-eleven translocation 1 gene protein homolog; ten-eleven translocation-1; tet oncogene 1; TET1 splice variant VP_DE4; TET1 splice variant VP_DE456

View more View less

Gene Aliases: 2510010B09Rik; AA517754; bA119F7.1; BB001228; CXXC6; D10Ertd17e; KIAA1676; LCX; mKIAA1676; MLL-TET1; TET1; TET1-MLL

View more View less

UniProt ID: (Human) Q8NFU7, (Mouse) Q3URK3

View more View less

Entrez Gene ID: (Human) 80312, (Mouse) 52463

View more View less

Function(s)
iron ion binding zinc ion binding structure-specific DNA binding methylcytosine dioxygenase activity DNA binding protein binding oxidoreductase activity metal ion binding dioxygenase activity
Process(es)
inner cell mass cell differentiation transcription, DNA-templated regulation of transcription, DNA-templated protein O-linked glycosylation chromatin modification stem cell population maintenance regulation of DNA methylation positive regulation of transcription from RNA polymerase II promoter oxidation-reduction process DNA demethylation negative regulation of methylation-dependent chromatin silencing regulation of genetic imprinting positive regulation of cell proliferation covalent chromatin modification positive regulation of histone methylation
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-c7d89468c-sbmwt:80/100.66.128.142:80.
git-commit: 4db89d68e0ee702f860d53b64def1eb94897fade
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0