Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • TUFT1 Antibodies

Abnova

TUFT1 Monoclonal Antibody (2C10)

View all (7) TUFT1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite TUFT1 Monoclonal Antibody (2C10)

  • Antibody Testing Data (1)
TUFT1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
TUFT1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

TUFT1 Antibody (H00007286-M01) in WB

Western Blot analysis of TUFT1 expression in transfected 293T cell line by TUFT1 monoclonal antibody (M01), clone 2C10. Lane 1: TUFT1 transfected lysate (Predicted MW: 44.3 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

TUFT1 Antibody in Western Blot (WB)
TUFT1 Monoclonal Antibody (2C10)

Product Details

H00007286-M01

Applications
Tested Dilution
Publications

Western Blot (WB)

1-5 µg/mL
-

ELISA (ELISA)

10 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2a, kappa

Class

Monoclonal

Type

Antibody

Clone

2C10

Immunogen

TUFT1 (NP_064512.1, 241 a.a. approximately 350 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.2 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Wet ice

Product Specific Information

Product may be used with Western Blot (Transfected lysate).

Immunogen sequence: TEHQALLAKV REGEVALEEL RSNNADCQAE REKAATLEKE VAGLREKIHH LDDMLKSQQR KVRQMIEQLQ NSKAVIQSKD ATIQELKEKI AYLEAENLEM HDRMEHLIEK

Target Information

TUFT1 is involved in the mineralization and structural organization of enamel.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Tuftelin; UNQ865

View more View less

Gene Aliases: TUFT1

View more View less

UniProt ID: (Human) Q9NNX1

View more View less

Entrez Gene ID: (Human) 7286

View more View less

Function(s)
protein binding structural constituent of tooth enamel
Process(es)
bone mineralization intracellular signal transduction odontogenesis
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0