Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • TRF2 Antibodies

Fabgennix

TRF2 Polyclonal Antibody, Biotin

View all (29) TRF2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite TRF2 Polyclonal Antibody, Biotin

TRF2 Polyclonal Antibody, Biotin

Product Details

TRF2-201-BIOTIN

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:1,000
-

ELISA (ELISA)

1:10,000
-

Immunoprecipitation (IP)

1:50-1:250
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Synthetic peptide taken within amino acid region 80-130 on human Telomeric repeat-binding factor 2 protein.
View immunogen

Conjugate

Biotin Biotin Biotin

Form

Liquid

Concentration

0.5-1.5 mg/mL

Purification

Affinity chromatography

Storage buffer

proprietary buffer, pH 7.4-7.8, with 30% glycerol, 0.5% BSA

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

TRF2 is a ubiquitously expressed protein that is implicated in the control of telomere length. TRF2, like TRF1 contains a Myb-related DNA binding motif. It binds to duplex TTAGGG repeats and is localized to all human telomeres in metaphase chromosomes. TRF2 is thought to protect chromosome ends by maintaining the correct structure at telomere termini. The use of mutant forms of TRF2 has implicated a role for TRF2 in the prevention of senescence in primary human cells. Recently, it has been shown that inhibition of TRF2 resulted in apoptosis in a subset of mammalian cell types.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Telomeric DNA-binding protein; telomeric repeat binding protein 2; Telomeric repeat-binding factor 2; TTAGGG repeat-binding factor 2

View more View less

Gene Aliases: TERF2; TRBF2; TRF2

View more View less

UniProt ID: (Human) Q15554, (Mouse) O35144

View more View less

Entrez Gene ID: (Human) 7014, (Rat) 361403, (Mouse) 21750

View more View less

Function(s)
double-stranded telomeric DNA binding telomerase activity protein binding protein C-terminus binding telomeric DNA binding protein homodimerization activity macromolecular complex binding G-rich strand telomeric DNA binding chromatin binding DNA binding
Process(es)
telomere maintenance in utero embryonic development positive regulation of gene expression negative regulation of gene expression obsolete telomere maintenance via telomere shortening telomere capping telomeric loop formation protection from non-homologous end joining at telomere negative regulation of telomere maintenance positive regulation of telomere maintenance positive regulation of nitric-oxide synthase activity protein localization to chromosome, telomeric region negative regulation of beta-galactosidase activity negative regulation of cellular senescence cell cycle regulation of telomere maintenance telomere maintenance via telomerase negative regulation of telomere maintenance via recombination regulation of telomere maintenance via telomerase negative regulation of telomere maintenance via semi-conservative replication cellular senescence negative regulation of telomere single strand break repair negative regulation of telomere capping negative regulation of t-circle formation
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0