Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • TACI Antibodies

Abnova

TNFRSF13B Monoclonal Antibody (1F5)

View all (41) TACI antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite TNFRSF13B Monoclonal Antibody (1F5)

TNFRSF13B Monoclonal Antibody (1F5)

Product Details

H00023495-M20

Applications
Tested Dilution
Publications

Western Blot (WB)

1-5 µg/mL
-

ELISA (ELISA)

10 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1, kappa

Class

Monoclonal

Type

Antibody

Clone

1F5

Immunogen

TNFRSF13B (NP_036584.1, 2 a.a. approximately 165 a.a) partial recombinant protein.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

See Label

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Sequence of this protein is as follows: SGLGRSRRGG RSRVDQEERF PQGLWTGVAM RSCPEEQYWD PLLGTCMSCK TICNHQSQRT CAAFCRSLSC RKEQGKFYDH LLRDCISCAS ICGQHPKQCA YFCENKLRSP VNLPPELRRQ RSGEVENNSD NSGRYQGLEH RGSEASPALP GLKLSADQVA LVYS

Target Information

Members in the TNF superfamily regulate immune responses and induce apoptosis. Two novel members in the TNF family were recently identified and designated BAFF/BLyS/TALL-1/THANK/zTNF4 and April/TALL-2, respectively. BAFF was characterized as a B cell activator since it induced B cell proliferation and immunoglobulin secretion. April regulates immunological and non-immunological cell growth. Three receptors, BCMA, TACI, and BAFF-R, for BAFF and April were recently identified. TACI, like BCMA, binds BAFF and April. TACI and its ligands regulate humoral immune responses, activate NF-kappa-B and c-jun N-terminal kinase, and are involved in B cell associated autoimmune diseases.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CD267; Transmembrane activator and CAML interactor; tumor necrosis factor receptor 13B; Tumor necrosis factor receptor superfamily member 13B; tumor necrosis factor receptor superfamily, member 13B

View more View less

Gene Aliases: CD267; CVID; CVID2; IGAD2; RYZN; TACI; TNFRSF13B; TNFRSF14B

View more View less

UniProt ID: (Human) O14836

View more View less

Entrez Gene ID: (Human) 23495

View more View less

Function(s)
receptor activity protein binding
Process(es)
B cell homeostasis hematopoietic progenitor cell differentiation adaptive immune response cell surface receptor signaling pathway negative regulation of B cell proliferation tumor necrosis factor-mediated signaling pathway
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0