Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • TIMP1 Antibodies

NeoBiotechnologies

TIMP1 (Colorectal and Lymph Node Metastasis Marker) Recombinant Rabbit Monoclonal Antibody (TIMP1/1944R)

View all (52) TIMP1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite TIMP1 (Colorectal and Lymph Node Metastasis Marker) Recombinant Rabbit Monoclonal Antibody (TIMP1/1944R)

TIMP1 (Colorectal and Lymph Node Metastasis Marker) Recombinant Rabbit Monoclonal Antibody (TIMP1/1944R)
  • Antibody Testing Data (1)
TIMP1 (Colorectal and Lymph Node Metastasis Marker) Antibody in SDS-PAGE (SDS-PAGE)
Group 53 Created with Sketch.
TIMP1 (Colorectal and Lymph Node Metastasis Marker) Antibody in SDS-PAGE (SDS-PAGE)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

TIMP1 (Colorectal and Lymph Node Metastasis Marker) Antibody (7076-RBM4-P1ABX) in SDS-PAGE

SDS-PAGE analysis of Purified TIMP1 Rabbit Recombinant Monoclonal Antibody (TIMP1/1944R). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

TIMP1 (Colorectal and Lymph Node Metastasis Marker) Antibody in SDS-PAGE (SDS-PAGE)

Product Details

7076-RBM4-P1ABX

Applications
Tested Dilution
Publications

ELISA (ELISA)

2-4 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

TIMP1/1944R

Immunogen

Recombinant human full-length TIMP1 protein
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

TIMP1 (metalloproteinase inhibitor 1) is a metalloproteinase inhibitor that functions by forms one-to-one complexes with target metallproteinases, such as collagenases, and irreversible inactivates them by binding to their catalytic zinc cofactor. TIMP1 acts on MMP1, MMP2, MMP3, MMP7, MMP8, MMP9, MMP10, MMP11, MMP12, MMP14, and MMP16. It also functions as a growth factor that regulates cell differenation, migration, and cell death. TIMP1 activates cellular signaling cascades via CD63 and ITGB1.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Collagenase inhibitor; EPA; erythroid potentiating activity; Erythroid-potentiating activity; Fibroblast collagenase inhibitor; FLJ90373; Metalloproteinase inhibitor; Metalloproteinase inhibitor 1; MMP inhibitor; RP1-230G1.3; TIMP 1; TIMP-1; Tissue inhibitor of metalloproteinases 1

View more View less

Gene Aliases: CLGI; EPA; EPO; HCI; TIMP; TIMP-1; TIMP1

View more View less

UniProt ID: (Human) P01033

View more View less

Entrez Gene ID: (Human) 7076

View more View less

Function(s)
protease binding cytokine activity protein binding growth factor activity metalloendopeptidase inhibitor activity metal ion binding
Process(es)
cell activation platelet degranulation aging positive regulation of cell proliferation response to hormone negative regulation of endopeptidase activity extracellular matrix disassembly response to cytokine wound healing negative regulation of apoptotic process negative regulation of catalytic activity response to peptide hormone negative regulation of membrane protein ectodomain proteolysis cartilage development negative regulation of trophoblast cell migration regulation of integrin-mediated signaling pathway
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5cc698dfdd-6b6sp:80/100.66.129.221:80.
git-commit: 7dd0da31084df89e35c5ffd4006b5f1c529b0b63
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0