Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • TIE1 Antibodies

Invitrogen

TIE1 Monoclonal Antibody (4E7F11)

View all (20) TIE1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite TIE1 Monoclonal Antibody (4E7F11)

TIE1 Monoclonal Antibody (4E7F11)

Product Details

MA5-30934

Applications
Tested Dilution
Publications

ELISA (ELISA)

1:250-1:2,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2b

Class

Monoclonal

Type

Antibody

Clone

4E7F11

Immunogen

Recombinant Human Tie1 Protein (Met1-Gln760)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

PBS

Contains

no preservative

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2786603

Product Specific Information

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).

This antibody has specificity for Human Tie1.

Target Information

Probable protein tyrosine-kinase transmembrane receptor.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: tyrosine kinase with immunoglobulin and epidermal growth factor homology domains 1; tyrosine kinase with immunoglobulin-like and EGF-like domains 1; Tyrosine-protein kinase receptor Tie-1

View more View less

Gene Aliases: JTK14; TIE; TIE1

View more View less

UniProt ID: (Human) P35590

View more View less

Entrez Gene ID: (Human) 7075

View more View less

Function(s)
transmembrane receptor protein tyrosine kinase activity protein binding ATP binding
Process(es)
angiogenesis vasculogenesis in utero embryonic development signal transduction mesoderm development negative regulation of angiogenesis peptidyl-tyrosine phosphorylation negative regulation of cell migration response to retinoic acid plasma membrane fusion
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0