Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: TLMTTDCPEP LLQPWLEGWH VEGTDRLRVS WSLPLVPGPL VGDGFLLRLW DGTRGQERRE NVSSPQARTA LLTGLTPGTH YQLDVQLYHC TLLGPASPPA HVLLPPSG*
Probable protein tyrosine-kinase transmembrane receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: tyrosine kinase with immunoglobulin and epidermal growth factor homology domains 1; tyrosine kinase with immunoglobulin-like and EGF-like domains 1; Tyrosine-protein kinase receptor Tie-1
Gene Aliases: JTK14; TIE; TIE1
UniProt ID: (Human) P35590
Entrez Gene ID: (Human) 7075
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support