Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • TGFBR3 Antibodies

Bioss

TGF Beta R3 Polyclonal Antibody

View all (19) TGFBR3 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite TGF Beta R3 Polyclonal Antibody

  • Antibody Testing Data (1)
TGF Beta R3 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
TGF Beta R3 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

TGF Beta R3 Antibody (BS-1910R) in WB

Lane 1: K562 Cells; Lane 2: HepG2 Cells; Lane 3: Hela Cells; Probed with TGF Beta R3 Polyclonal Antibody (bs-1910R) at 1:300 overnight at 4°C followed by a conjugated secondary antibody for 60 minutes at 37°C. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

TGF Beta R3 Antibody in Western Blot (WB)
TGF Beta R3 Polyclonal Antibody

Product Details

BS-1910R

Applications
Tested Dilution
Publications

Western Blot (WB)

1:300
-

Immunohistochemistry (Paraffin) (IHC (P))

1:200-1:400
-

Immunohistochemistry (Frozen) (IHC (F))

1:100-1:500
-

Immunocytochemistry (ICC/IF)

1:50-1:200
-

ELISA (ELISA)

1:500-1:1,000
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

KLH conjugated synthetic peptide derived from human TGFBR3, amino acids 701-850.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

0.01M TBS, pH 7.4, with 1% BSA, 50% glycerol

Contains

0.02% ProClin 300

Storage conditions

-20°C

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

This locus encodes the transforming growth factor (TGF)-beta type III receptor. The encoded receptor is a membrane proteoglycan that often functions as a co-receptor with other TGF-beta receptor superfamily members. Ectodomain shedding produces soluble TGFBR3, which may inhibit TGFB signaling. Decreased expression of this receptor has been observed in various cancers. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Betaglycan; betaglycan proteoglycan; OTTHUMP00000011787; tgf beta receptor 3; TGF-beta receptor type 3; TGF-beta receptor type III; TGFR-3; Transforming growth factor beta receptor III; Transforming growth factor beta receptor type 3; transforming growth factor, beta receptor 3

View more View less

Gene Aliases: 1110036H20Rik; AU015626; AW215636; betaglycan; BGCAN; TBRIII; TGFBR3

View more View less

UniProt ID: (Human) Q03167, (Mouse) O88393, (Rat) P26342

View more View less

Entrez Gene ID: (Human) 7049, (Mouse) 21814, (Rat) 29610

View more View less

Function(s)
transforming growth factor beta-activated receptor activity type II transforming growth factor beta receptor binding transforming growth factor beta receptor binding protein binding glycosaminoglycan binding heparin binding coreceptor activity fibroblast growth factor binding PDZ domain binding SMAD binding activin binding transforming growth factor beta binding transforming growth factor beta receptor activity, type III
Process(es)
blood vessel development vasculogenesis osteoblast differentiation in utero embryonic development blastocyst development epithelial to mesenchymal transition liver development blood vessel remodeling epicardial cell to mesenchymal cell transition protein complex assembly protein phosphorylation immune response transforming growth factor beta receptor signaling pathway transforming growth factor beta receptor complex assembly positive regulation of cell proliferation embryo development negative regulation of epithelial cell migration negative regulation of epithelial to mesenchymal transition cell growth BMP signaling pathway positive regulation of transforming growth factor beta receptor signaling pathway negative regulation of transforming growth factor beta receptor signaling pathway positive regulation of BMP signaling pathway response to follicle-stimulating hormone response to prostaglandin E response to luteinizing hormone intracellular signal transduction regulation of protein binding negative regulation of epithelial cell proliferation regulation of blood vessel size positive regulation of NF-kappaB transcription factor activity negative regulation of cellular component movement ventricular cardiac muscle tissue morphogenesis palate development cardiac muscle cell proliferation definitive hemopoiesis cardiac epithelial to mesenchymal transition definitive erythrocyte differentiation heart trabecula formation pathway-restricted SMAD protein phosphorylation coronary vasculature development coronary vasculature morphogenesis vasculogenesis involved in coronary vascular morphogenesis visceral serous pericardium development negative regulation of apoptotic process involved in morphogenesis response to hypoxia signal transduction organ regeneration heart morphogenesis cell migration
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0