Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • TERT Antibodies

Osenses

TERT Polyclonal Antibody

2 References
View all (37) TERT antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite TERT Polyclonal Antibody

TERT Polyclonal Antibody

Product Details

OST00412W-100UL

Applications
Tested Dilution
Publications

Western Blot (WB)

1:300-1:2,000
-

Immunohistochemistry (IHC)

1:300-1:2,000
-

Miscellaneous PubMed (MISC)

-
View 2 publications 2 publications
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / Ig

Class

Polyclonal

Type

Antibody

Immunogen

A synthetic peptide from aa region 950-1000 of human TERT conjugated to an immunogenic carrier protein was used as the antigen
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

Conc. Not Determined

Storage buffer

whole serum

Contains

no preservative

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.

Specificity of this antibody: TERT.

Target Information

Telomerase is a ribonucleoprotein polymerase that maintains telomere ends by addition of the telomere repeat TTAGGG. The enzyme consists of a protein component with reverse transcriptase activity, encoded by this gene, and an RNA component which serves as a template for the telomere repeat. Telomerase expression plays a role in cellular senescence, as it is normally repressed in postnatal somatic cells resulting in progressive shortening of telomeres. Deregulation of telomerase expression in somatic cells may be involved in oncogenesis. Studies in mouse suggest that telomerase also participates in chromosomal repair, since de novo synthesis of telomere repeats may occur at double-stranded breaks. Alternatively spliced variants encoding different isoforms of telomerase reverse transcriptase (TERT) have been identified; the full-length sequence of some variants has not been determined. Alternative splicing at this locus is thought to be one mechanism of regulation of telomerase activity.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: HEST2; hTERT; Telomerase catalytic subunit; Telomerase reverse transcriptase; Telomerase-associated protein 2; TP2

View more View less

Gene Aliases: CMM9; DKCA2; DKCB4; EST2; hEST2; hTRT; PFBMFT1; TCS1; TERT; TP2; TRT

View more View less

UniProt ID: (Human) O14746

View more View less

Entrez Gene ID: (Human) 7015

View more View less

Function(s)
tRNA binding transcription coactivator binding DNA binding telomerase activity telomerase RNA reverse transcriptase activity RNA binding RNA-directed DNA polymerase activity RNA-directed RNA polymerase activity protein binding nucleotidyltransferase activity telomeric DNA binding protein homodimerization activity metal ion binding telomerase RNA binding
Process(es)
telomere maintenance transcription, RNA-templated RNA-dependent DNA biosynthetic process telomere maintenance via telomerase mitochondrion organization negative regulation of gene expression DNA strand elongation positive regulation of Wnt signaling pathway production of siRNA involved in RNA interference regulation of protein stability positive regulation of protein binding RNA biosynthetic process positive regulation of hair cycle positive regulation of nitric-oxide synthase activity establishment of protein localization to telomere cellular response to hypoxia DNA biosynthetic process replicative senescence positive regulation of pri-miRNA transcription from RNA polymerase II promoter negative regulation of production of siRNA involved in RNA interference positive regulation of protein localization to nucleolus beta-catenin-TCF complex assembly positive regulation of stem cell proliferation negative regulation of cellular senescence negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0