Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD248 Antibodies

Proteintech

TEM1 Monoclonal Antibody (1F9B4)

11 References
View all (16) CD248 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite TEM1 Monoclonal Antibody (1F9B4)

  • Antibody Testing Data (6)
TEM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
TEM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 6

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

TEM1 Antibody (60170-1-IG) in IHC (P)

Immunohistochemistry of paraffin-embedded human renal cell carcinoma tissue slide using 60170-1-Ig (TEM1 antibody at dilution of 1:250 (under 40x lens). Heat mediated antigen retrieved with Tris-EDTA buffer (pH 9). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

TEM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
TEM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
TEM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
TEM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
TEM1 Antibody in Western Blot (WB)
TEM1 Antibody in Western Blot (WB)
TEM1 Monoclonal Antibody (1F9B4)

Product Details

60170-1-IG

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000-1:4,000
View 4 publications 4 publications

Immunohistochemistry (IHC)

-
View 2 publications 2 publications

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:500
-

Immunocytochemistry (ICC/IF)

-
View 3 publications 3 publications

Immunoprecipitation (IP)

-
View 2 publications 2 publications
Product Specifications

Species Reactivity

Human, Mouse

Published species

Human, Mouse, Rat

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

1F9B4

Immunogen

TEM1 Fusion Protein Ag13334 (399-687 aa encoded by BC051340)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Caprylic acid precipitation

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice (domestic); Dry ice (international)

Product Specific Information

Immunogen sequence: EPTQPPDFA LAYRPSFPED REPQIPYPEP TWPPPLSAPR VPYHSSVLSV TRPVVVSATH PTLPSAHQPP VIPATHPALS RDHQIPVIAA NYPDLPSAYQ PGILSVSHSA QPPAHQPPMI STKYPELFPA HQSPMFPDTR VAGTQTTTHL PGIPPNHAPL VTTLGAQLPP QAPDALVLRT QATQLPIIPT AQPSLTTTSR SPVSPAHQIS VPAATQPAAL PTLLPSQSPT NQTSPISPTH PHSKAPQIPR EDGPSPKLAL WLPSPAPTAA PTALGEAGLA EHSQRDDRWL (399-687 aa encoded by BC051340)

Target Information

CD248 (CD248 Molecule) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include calcium ion binding and extracellular matrix binding. An important paralog of this gene is CD93. CD248 may play a role in tumor angiogenesis.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: 2610111G01Rik; CD164 sialomucin-like 1; CD248; CD248 antigen, endosialin; CD248 molecule, endosialin; Endosialin; MGC119478; MGC119479; Tumor endothelial marker 1

View more View less

Gene Aliases: 2610111G01Rik; AI842296; CD164L1; CD248; TEM1

View more View less

UniProt ID: (Human) Q9HCU0, (Mouse) Q91V98

View more View less

Entrez Gene ID: (Human) 57124, (Mouse) 70445

View more View less

Function(s)
molecular_function calcium ion binding carbohydrate binding extracellular matrix binding
Process(es)
positive regulation of cell proliferation cell migration lymph node development anatomical structure regression positive regulation of endothelial cell apoptotic process biological_process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0