Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • TAF11 Antibodies

Abnova

TAF11 Monoclonal Antibody (3D3)

View all (12) TAF11 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite TAF11 Monoclonal Antibody (3D3)

  • Antibody Testing Data (4)
TAF11 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
TAF11 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

TAF11 Antibody (H00006882-M01) in ICC/IF

Immunofluorescence of monoclonal antibody to TAF11 on HeLa cell. Antibody concentration is 10 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

TAF11 Antibody in Immunocytochemistry (ICC/IF)
TAF11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
TAF11 Antibody in Western Blot (WB)
TAF11 Antibody in ELISA (ELISA)
TAF11 Monoclonal Antibody (3D3)

Product Details

H00006882-M01

Applications
Tested Dilution
Publications

Western Blot (WB)

1-5 µg/mL
-

Immunohistochemistry (Paraffin) (IHC (P))

3 µg/mL
-

Immunocytochemistry (ICC/IF)

10 µg/mL
-

ELISA (ELISA)

0.1 ng/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1, kappa

Class

Monoclonal

Type

Antibody

Clone

3D3

Immunogen

TAF11 (NP_005634, 158 a.a. approximately 210 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

See Label

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Sequence of this protein is as follows: SKVFVGEVVE EALDVCEKWG EMPPLQPKHM REAVRRLKSK GQIPNSKHKK IIF

Target Information

TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. TAF11 is a small subunit of TFIID that is present in all TFIID complexes and interacts with TBP. This subunit also interacts with another small subunit, TAF13, to form a heterodimer with a structure similar to the histone core structure. Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: TAF(II)28; TAF11 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 28kDa; TAFII-28; TATA box binding protein (TBP)-associated factor, RNA polymerase II, I, 28kD; TATA box binding protein associated factor 11; TFIID subunit p30-beta; transcription initiation factor TFIID 28 kD subunit; Transcription initiation factor TFIID 28 kDa subunit; Transcription initiation factor TFIID subunit 11

View more View less

Gene Aliases: MGC:15243; PRO2134; TAF11; TAF2I; TAFII28

View more View less

UniProt ID: (Human) Q15544

View more View less

Entrez Gene ID: (Human) 6882

View more View less

Function(s)
transcription coactivator activity protein binding vitamin D receptor binding thyroid hormone receptor binding protein heterodimerization activity protein N-terminus binding
Process(es)
regulation of transcription, DNA-templated transcription from RNA polymerase II promoter transcription initiation from RNA polymerase II promoter transcription elongation from RNA polymerase II promoter snRNA transcription from RNA polymerase II promoter positive regulation by host of viral transcription RNA polymerase II transcriptional preinitiation complex assembly regulation of signal transduction by p53 class mediator
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0