Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Synuclein alpha/beta Antibodies

Invitrogen

Synuclein alpha/beta Recombinant Rabbit Monoclonal Antibody (9E9X5)

View all (5) Synuclein alpha/beta antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Synuclein alpha/beta Recombinant Rabbit Monoclonal Antibody (9E9X5)

  • Antibody Testing Data (3)
Synuclein alpha/beta Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
Synuclein alpha/beta Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Synuclein alpha/beta Antibody (MA5-35604) in IHC (P)

Immunohistochemistry (Immunofluorescence) analysis of Synuclein alpha/beta in rat brain. Samples were incubated with Synuclein alpha/beta Monoclonal antibody (Product # MA5-35604) using a dilution of 1:100 (40x lens). Blue: DAPI for nuclear staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Synuclein alpha/beta Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Synuclein alpha/beta Antibody in Western Blot (WB)
Synuclein alpha/beta Antibody in Western Blot (WB)
Synuclein alpha/beta Recombinant Rabbit Monoclonal Antibody (9E9X5)

Product Details

MA5-35604

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:2,000
-

Immunohistochemistry (Paraffin) (IHC (P))

Assay-dependent
-

Immunocytochemistry (ICC/IF)

1:50-1:200
-

ELISA (ELISA)

1 µg/mL
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

9E9X5

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human alpha + beta Synuclein (P37840)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1.35 mg/mL

Purification

Affinity Chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol, 0.05% BSA

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2849504

Product Specific Information

Immunogen sequence: MDVFMKGLSK AKEGVVAAAE KTKQGVAEAA GKTKEGVLYV GSKTKEGVVH GVATVAEKTK EQVTNVGGAV VTGVTAVAQK TVEGAGSIAA ATGFVKKDQL

Target Information

Synuclein family of proteins is abundantly expressed in neuronal cytosol and presynaptic terminals. In vertebrates they are encoded by three different genes. Synucleins have been specifically implemented in three major diseases: Alzheimer's (AD), Parkinson's PD) and breast cancer. In AD, a peptide derived from alpha synuclein forms an intrinsic component of plaque amyloid. In PD, an alpha synuclein accumulates in Lewy bodies. An allele of alpha synuclein has been linked to many familial cases of PD. In breast cancer increased expression of gamma synuclein correlates with the disease progression. Synucleins appear to be involved in the membrane plasticity in developing song control system of songbirds.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Alpha-synuclein; Beta-synuclein; NACP; non A-beta component of AD amyloid; Non-A beta component of AD amyloid; non-A4 component of amyloid; Non-A4 component of amyloid precursor; Phosphoneuroprotein 14; PNP 14; synuclein alpha-140; synuclein, alpha (non A4 component of amyloid precursor)

View more View less

Gene Aliases: AI838531; alphaSYN; betaSYN; NACP; PARK1; PARK4; PD1; SNCA; SNCB; Syn

View more View less

UniProt ID: (Human) P37840, (Human) Q16143, (Rat) P37377, (Mouse) O55042, (Mouse) Q91ZZ3, (Rat) Q63754

View more View less

Entrez Gene ID: (Human) 6622, (Human) 6620, (Rat) 29219, (Mouse) 20617, (Mouse) 104069, (Rat) 113893

View more View less

Function(s)
magnesium ion binding fatty acid binding copper ion binding calcium ion binding protein binding phospholipid binding microtubule binding ferrous iron binding zinc ion binding oxidoreductase activity kinesin binding protein domain specific binding histone binding identical protein binding alpha-tubulin binding cysteine-type endopeptidase inhibitor activity involved in apoptotic process phospholipase binding transcription regulatory region DNA binding dynein binding protein N-terminus binding tau protein binding beta-tubulin binding phosphoprotein binding phospholipase D inhibitor activity phospholipase inhibitor activity enzyme binding Hsp70 protein binding arachidonic acid binding metal ion binding
Process(es)
negative regulation of transcription from RNA polymerase II promoter microglial cell activation positive regulation of receptor recycling negative regulation of protein phosphorylation positive regulation of neurotransmitter secretion fatty acid metabolic process neutral lipid metabolic process phospholipid metabolic process apoptotic process activation of cysteine-type endopeptidase activity involved in apoptotic process mitochondrial membrane organization aging adult locomotory behavior response to iron(II) ion regulation of phospholipase activity negative regulation of platelet-derived growth factor receptor signaling pathway regulation of glutamate secretion regulation of dopamine secretion negative regulation of microtubule polymerization receptor internalization protein destabilization response to magnesium ion negative regulation of transporter activity response to lipopolysaccharide negative regulation of monooxygenase activity positive regulation of peptidyl-serine phosphorylation response to interferon-gamma cellular response to oxidative stress negative regulation of histone acetylation regulation of locomotion dopamine biosynthetic process response to drug mitochondrial ATP synthesis coupled electron transport regulation of macrophage activation positive regulation of apoptotic process negative regulation of apoptotic process negative regulation of cysteine-type endopeptidase activity involved in apoptotic process extracellular fibril organization negative regulation of neuron apoptotic process cellular protein metabolic process cellular response to fibroblast growth factor stimulus positive regulation of endocytosis negative regulation of exocytosis negative regulation of dopamine metabolic process behavioral response to cocaine regulation of long-term neuronal synaptic plasticity synaptic vesicle endocytosis synapse organization regulation of acyl-CoA biosynthetic process positive regulation of release of sequestered calcium ion into cytosol dopamine uptake involved in synaptic transmission negative regulation of dopamine uptake involved in synaptic transmission negative regulation of serotonin uptake negative regulation of norepinephrine uptake calcium ion homeostasis oxidation-reduction process excitatory postsynaptic potential long-term synaptic potentiation positive regulation of inositol phosphate biosynthetic process negative regulation of thrombin receptor signaling pathway response to interleukin-1 cellular response to copper ion cellular response to epinephrine stimulus positive regulation of protein serine/threonine kinase activity negative regulation of neuron death negative regulation of mitochondrial electron transport, NADH to ubiquinone positive regulation of glutathione peroxidase activity positive regulation of hydrogen peroxide catabolic process regulation of synaptic vesicle recycling regulation of reactive oxygen species biosynthetic process negative regulation of chaperone-mediated autophagy synaptic transmission, dopaminergic chemical synaptic transmission response to cocaine dopamine metabolic process regulation of neuron apoptotic process regulation of neurotransmitter secretion regulation of neuronal synaptic plasticity synaptic vesicle transport positive regulation of synaptic transmission membrane organization regulation of neuron death regulation of reactive oxygen species metabolic process negative regulation of catalytic activity
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0