Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • SULT1C3 Antibodies

Abnova

SULT1C3 Polyclonal Antibody, MaxPab™

View all (2) SULT1C3 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite SULT1C3 Polyclonal Antibody, MaxPab™

  • Antibody Testing Data (1)
SULT1C3 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
SULT1C3 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

SULT1C3 Antibody (H00442038-B01P) in WB

Western Blot analysis of SULT1C3 expression in transfected 293T cell line (H00442038-T01) by SULT1C3 MaxPab polyclonal antibody.

Lane 1: SULT1C3 transfected lysate(33.44 KDa).
Lane 2: Non-transfected lysate.
{{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

SULT1C3 Antibody in Western Blot (WB)
SULT1C3 Polyclonal Antibody, MaxPab™

Product Details

H00442038-B01P

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:1,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG

Class

Polyclonal

Type

Antibody

Immunogen

SULT1C3 (AAI46363.1, 1 a.a. approximately 304 a.a) full-length human protein.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

See Label

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Sequence of this protein is as follows: MAKIEKNAPT MEKKPELFNI MEVDGVPTLI LSKEWWEKVC NFQAKPDDLI LATYPKSGTT WMHEILDMIL NDGDVEKCKR AQTLDRHAFL ELKFPHKEKP DLEFVLEMSS PQLIKTHLPS HLIPPSIWKE NCKIVYVARN PKDCLVSYYH FHRMASFMPD PQNLEEFYEK FMSGKVVGGS WFDHVKGWWA AKDMHRILYL FYEDIKKDPK REIEKILKFL EKDISEEILN KIIYHTSFDV MKQNPMTNYT TLPTSIMDHS ISPFMRKGMP GDWKNYFTVA QNEEFDKDYQ KKMAGSTLTF RTEI

Target Information

Has low sulphotransferase activity towards various substrates with alcohol groups (in vitro). May catalyze the sulfate conjugation of xenobiotic compounds and endogenous substrates.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: ST1C3; Sulfotransferase 1C3; sulfotransferase family, cytosolic, 1C, member 3

View more View less

Gene Aliases: ST1C3; SULT1C3

View more View less

UniProt ID: (Human) Q6IMI6

View more View less

Entrez Gene ID: (Human) 442038

View more View less

Function(s)
alcohol sulfotransferase activity aryl sulfotransferase activity
Process(es)
sulfur compound metabolic process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0