Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • STIL Antibodies

Proteintech

STIL Monoclonal Antibody (1E12C1)

3 References
View all (8) STIL antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite STIL Monoclonal Antibody (1E12C1)

  • Antibody Testing Data (3)
STIL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
STIL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

STIL Antibody (66876-1-IG) in IHC (P)

Immunohistochemical analysis of paraffin-embedded human colon cancer tissue slide using 66876-1-Ig (STIL antibody) at dilution of 1:500 (under 40x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

STIL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
STIL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
STIL Antibody in Western Blot (WB)

Product Details

66876-1-IG

Applications
Tested Dilution
Publications

Western Blot (WB)

1:2,000
View 2 publications 2 publications

Immunohistochemistry (IHC)

-
View 1 publication 1 publication

Immunohistochemistry (Paraffin) (IHC (P))

1:500
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

1E12C1

Immunogen

STIL Fusion Protein Ag19124

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1000 µg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice

Product Specific Information

Aliquoting is unnecessary for -20°C storage.

Target Information

Plays an essential role in early embryonic development.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: SCL-interrupting locus protein; TAL-1-interrupting locus protein

View more View less

Gene Aliases: MCPH7; SIL; STIL

View more View less

UniProt ID: (Human) Q15468

View more View less

Entrez Gene ID: (Human) 6491

View more View less

Function(s)
protein binding
Process(es)
embryonic axis specification in utero embryonic development neural tube closure heart looping mitotic spindle organization smoothened signaling pathway determination of left/right symmetry cell proliferation neural tube development forebrain development notochord development floor plate development multicellular organism growth negative regulation of apoptotic process regulation of centriole replication centrosome duplication protein localization to centrosome
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0