Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • TRAP alpha Antibodies

OriGene

SSR1 Monoclonal Antibody (OTI4C7), TrueMAB™

Advanced Verification
View all (28) TRAP alpha antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite SSR1 Monoclonal Antibody (OTI4C7), TrueMAB™

  • Antibody Testing Data (15)
  • Advanced Verification (1)
SSR1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
SSR1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 16

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

SSR1 Antibody (CF503268) in ICC/IF

Anti-SSR1 mouse monoclonal antibody (TA503268) Immunofluorescent staining of COS7 cells transiently transfected by pCMV6-ENTRY SSR1(RC202408). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

SSR1 Antibody in Immunocytochemistry (ICC/IF)
SSR1 Antibody in Immunocytochemistry (ICC/IF)
SSR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
SSR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
SSR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
SSR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
SSR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
SSR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
SSR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
SSR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
SSR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
SSR1 Antibody in Western Blot (WB)
SSR1 Antibody in Western Blot (WB)
SSR1 Antibody in Western Blot (WB)
SSR1 Antibody in Flow Cytometry (Flow)
SSR1 Antibody
SSR1 Monoclonal Antibody (OTI4C7), TrueMAB™

Product Details

CF503268

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:2,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:150
-

Immunocytochemistry (ICC/IF)

1:50-1:100
-

Flow Cytometry (Flow)

1:100
-
Product Specifications

Species Reactivity

Dog, Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

OTI4C7

Immunogen

Full length human recombinant protein of human SSR1 produced in HEK293 cell.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.3, with 8% trehalose

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)

Target Information

The signal sequence receptor (SSR) is a glycosylated endoplasmic reticulum (ER) membrane receptor associated with protein translocation across the ER membrane. The SSR consists of 2 subunits, a 34-kD glycoprotein encoded by this gene and a 22-kD glycoprotein. This gene generates several mRNA species as a result of complex alternative polyadenylation. This gene is unusual in that it utilizes arrays of polyA signal sequences that are exclusively non-canonical.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: DKFZp781N23103; PGP35; Signal sequence receptor subunit alpha; signal sequence receptor, alpha; signal sequence receptor, alpha (translocon-associated protein alpha); SSR alpha subunit; SSR-alpha; SSRA precursor protein; translocon-associated protein alpha subunit; Translocon-associated protein subunit alpha; TRAP alpha; TRAP-alpha

View more View less

Gene Aliases: PSEC0262; SSR1; TRAPA

View more View less

UniProt ID: (Dog) P16967, (Human) P43307

View more View less

Entrez Gene ID: (Dog) 403951, (Human) 6745

View more View less

Function(s)
protein binding
Process(es)
cotranslational protein targeting to membrane positive regulation of cell proliferation IRE1-mediated unfolded protein response
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0