Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Syntrophin alpha-1 Antibodies

OriGene

SNTA1 Monoclonal Antibody (OTI2B4), TrueMAB™

View all (36) Syntrophin alpha-1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite SNTA1 Monoclonal Antibody (OTI2B4), TrueMAB™

  • Antibody Testing Data (3)
SNTA1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
SNTA1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

SNTA1 Antibody (CF502057) in ICC/IF

Anti-SNTA1 mouse monoclonal antibody (TA502057) Immunofluorescent staining of COS7 cells transiently transfected by pCMV6-ENTRY SNTA1(RC204480). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

SNTA1 Antibody in Immunocytochemistry (ICC/IF)
SNTA1 Antibody in Western Blot (WB)
SNTA1 Antibody in Flow Cytometry (Flow)
SNTA1 Monoclonal Antibody (OTI2B4), TrueMAB™

Product Details

CF502057

Applications
Tested Dilution
Publications

Western Blot (WB)

1:2,000
-

Immunocytochemistry (ICC/IF)

1:100
-

Flow Cytometry (Flow)

1:100
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

OTI2B4

Immunogen

Full length human recombinant protein of human SNTA1 produced in HEK293T cell.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.3, with 8% trehalose

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)

Target Information

Syntrophin alpha-1 (Dystrophin, SNTA1) is a large, rod-like cytoskeletal protein found at the inner surface of muscle fibers. Dystrophin is missing in Duchenne Muscular Dystrophy patients and is present in reduced amounts in Becker Muscular Dystrophy patients. The protein encoded by this gene is a peripheral membrane protein found associated with dystrophin and dystrophin-related proteins. This gene is a member of the syntrophin gene family, which contains at least two other structurally-related genes.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 59 kDa dystrophin-associated protein A1 acidic component 1; acidic alpha 1 syntrophin; Alpha-1-syntrophin; dystrophin-associated protein A1, 59kDa, acidic component; Pro-TGF-alpha cytoplasmic domain-interacting protein 1; RP23-192L3.1; syntrophin, alpha 1 (dystrophin-associated protein A1, 59kDa, acidic component); Syntrophin-1; TACIP1

View more View less

Gene Aliases: dJ1187J4.5; LQT12; SNT1; SNTA1; TACIP1

View more View less

UniProt ID: (Human) Q13424

View more View less

Entrez Gene ID: (Human) 6640

View more View less

Function(s)
actin binding structural molecule activity protein binding calmodulin binding sodium channel regulator activity PDZ domain binding ion channel binding nitric-oxide synthase binding ATPase binding
Process(es)
regulation of heart rate muscle contraction regulation of ventricular cardiac muscle cell membrane repolarization ventricular cardiac muscle cell action potential negative regulation of peptidyl-cysteine S-nitrosylation regulation of sodium ion transmembrane transport
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0