Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • SIRT1 Antibodies

Bioss

SIRT1/sirtuin 1 Polyclonal Antibody

View all (55) SIRT1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite SIRT1/sirtuin 1 Polyclonal Antibody

  • Antibody Testing Data (2)
SIRT1/sirtuin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
SIRT1/sirtuin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

SIRT1/sirtuin 1 Antibody (BS-2257R) in IHC (P)

Paraformaldehyde-fixed, paraffin embedded Mouse brain; Antigen retrieval by boiling in sodium citrate buffer (pH6.0) for 15min; Block endogenous peroxidase by 3% hydrogen peroxide for 20 minutes; Blocking buffer (normal goat serum) at 37°C for 30min; Antibody incubation with SIRT1/sirtuin 1 Polyclonal Antibody, Unconjugated (bs-2257R) at 1:400 overnight at 4°C, DAB staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

SIRT1/sirtuin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
SIRT1/sirtuin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
SIRT1/sirtuin 1 Polyclonal Antibody

Product Details

BS-2257R

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:2,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:400
-

Immunohistochemistry (Frozen) (IHC (F))

1:100-1:500
-

Immunocytochemistry (ICC/IF)

1:50-1:200
-

Flow Cytometry (Flow)

1:20-1:100
-

ELISA (ELISA)

1:500-1:1,000
-
Product Specifications

Species Reactivity

Dog, Human, Mouse

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

KLH conjugated synthetic peptide derived from human SIRT1, amino acids 551-650.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

0.01M TBS, pH 7.4, with 50% glycerol, 1% BSA

Contains

0.02% ProClin 300

Storage conditions

-20°C

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

NAD-dependent protein deacetylase sirtuin-1 (SIRT1) links transcriptional regulation directly to intracellular energetics and participates in the coordination of several separated cellular functions such as cell cycle, repsonse to DNA damage, metabolism, apoptosis, and autophagy. SIRT1 can modulate chromatin function through deacetylation of histones and can promote alterations in the methylation of histone and DNA, leading to transcriptional repression. It is essential in skeletal muscle cell differentiation and in response to low nutrients mediates the inhibitory effect on skeletal myoblast differentiation which also involves 5'-AMP-activated protein kinase (AMPK) and nicotinamide phosphoribosyltransferase (NAMPT).

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: hSIR2; hSIRT1; mSIR2a; NAD-dependent deacetylase sirtuin-1; NAD-dependent protein deacetylase sirtuin-1; NAD-dependent protein deacylase sirtuin-1; OTTHUMP00000198111; OTTHUMP00000198112; Regulatory protein SIR2 homolog 1; RP11-57G10.3; Sir2; sir2-like 1; SIR2-like protein 1; SIR2alpha; SIRT 1; SIRT-1; sirtuin 1 ((silent mating type information regulation 2, homolog) 1; sirtuin type 1

View more View less

Gene Aliases: AA673258; SIR2; Sir2a; SIR2alpha; SIR2L1; SIRT1

View more View less

UniProt ID: (Human) Q96EB6, (Mouse) Q923E4

View more View less

Entrez Gene ID: (Human) 23411, (Dog) 489012, (Mouse) 93759

View more View less

Function(s)
core promoter sequence-specific DNA binding p53 binding transcription corepressor activity NAD+ ADP-ribosyltransferase activity histone deacetylase activity protein binding protein C-terminus binding transcription factor binding NAD-dependent histone deacetylase activity deacetylase activity enzyme binding protein deacetylase activity NAD-dependent protein deacetylase activity nuclear hormone receptor binding histone binding identical protein binding HLH domain binding bHLH transcription factor binding metal ion binding NAD-dependent histone deacetylase activity (H3-K9 specific) mitogen-activated protein kinase binding NAD+ binding keratin filament binding protein-N-terminal asparagine amidohydrolase activity UDP-3-O-[3-hydroxymyristoyl] N-acetylglucosamine deacetylase activity hydrolase activity iprodione amidohydrolase activity (3,5-dichlorophenylurea)acetate amidohydrolase activity protein domain specific binding 4'-(2-hydroxyisopropyl)phenylurea amidohydrolase activity didemethylisoproturon amidohydrolase activity N-isopropylacetanilide amidohydrolase activity N-cyclohexylformamide amidohydrolase activity isonicotinic acid hydrazide hydrolase activity cis-aconitamide amidase activity gamma-N-formylaminovinylacetate hydrolase activity protein kinase B binding N2-acetyl-L-lysine deacetylase activity O-succinylbenzoate synthase activity indoleacetamide hydrolase activity N-acetylcitrulline deacetylase activity N-acetylgalactosamine-6-phosphate deacetylase activity diacetylchitobiose deacetylase activity chitooligosaccharide deacetylase activity
Process(es)
single strand break repair negative regulation of transcription from RNA polymerase II promoter chromatin silencing at rDNA pyrimidine dimer repair by nucleotide-excision repair DNA synthesis involved in DNA repair angiogenesis ovulation from ovarian follicle cellular glucose homeostasis positive regulation of protein phosphorylation positive regulation of endothelial cell proliferation positive regulation of adaptive immune response DNA replication DNA repair chromatin organization chromatin silencing establishment of chromatin silencing maintenance of chromatin silencing methylation-dependent chromatin silencing transcription, DNA-templated rRNA processing protein ADP-ribosylation protein deacetylation triglyceride mobilization cellular response to DNA damage stimulus response to oxidative stress spermatogenesis regulation of mitotic cell cycle muscle organ development cell aging positive regulation of cell proliferation cellular response to starvation negative regulation of gene expression positive regulation of cholesterol efflux regulation of lipid storage regulation of glucose metabolic process macrophage cytokine production positive regulation of phosphatidylinositol 3-kinase signaling viral process positive regulation of macroautophagy protein ubiquitination histone deacetylation peptidyl-lysine acetylation macrophage differentiation negative regulation of cell growth negative regulation of transforming growth factor beta receptor signaling pathway negative regulation of prostaglandin biosynthetic process protein destabilization positive regulation of chromatin silencing negative regulation of TOR signaling regulation of endodeoxyribonuclease activity negative regulation of NF-kappaB transcription factor activity response to insulin circadian regulation of gene expression regulation of protein import into nucleus, translocation leptin-mediated signaling pathway regulation of smooth muscle cell apoptotic process peptidyl-lysine deacetylation cellular triglyceride homeostasis regulation of peroxisome proliferator activated receptor signaling pathway regulation of cell proliferation negative regulation of phosphorylation response to hydrogen peroxide behavioral response to starvation cholesterol homeostasis intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator positive regulation of apoptotic process negative regulation of apoptotic process negative regulation of I-kappaB kinase/NF-kappaB signaling proteasome-mediated ubiquitin-dependent protein catabolic process positive regulation of cysteine-type endopeptidase activity involved in apoptotic process negative regulation of sequence-specific DNA binding transcription factor activity negative regulation of DNA damage response, signal transduction by p53 class mediator response to leptin positive regulation of MHC class II biosynthetic process negative regulation of fat cell differentiation positive regulation of DNA repair positive regulation of angiogenesis negative regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of insulin receptor signaling pathway white fat cell differentiation negative regulation of helicase activity positive regulation of histone H3-K9 methylation negative regulation of protein kinase B signaling fatty acid homeostasis negative regulation of androgen receptor signaling pathway histone H3-K9 modification cellular response to hydrogen peroxide regulation of bile acid biosynthetic process UV-damage excision repair histone H3 deacetylation cellular response to tumor necrosis factor negative regulation of histone H3-K14 acetylation cellular response to hypoxia cellular response to ionizing radiation regulation of protein serine/threonine kinase activity regulation of brown fat cell differentiation stress-induced premature senescence regulation of cellular response to heat negative regulation of neuron death negative regulation of protein acetylation negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway positive regulation of adipose tissue development histone H3-K9 deacetylation positive regulation of macrophage apoptotic process negative regulation of cAMP-dependent protein kinase activity positive regulation of cAMP-dependent protein kinase activity negative regulation of histone H4-K16 acetylation negative regulation of cellular response to testosterone stimulus negative regulation of peptidyl-lysine acetylation negative regulation of cellular senescence positive regulation of cellular senescence regulation of transcription, DNA-templated apoptotic process multicellular organism development intrinsic apoptotic signaling pathway in response to DNA damage positive regulation of heart rate negative regulation of cardiac muscle cell apoptotic process positive regulation of neuron projection development positive regulation of skeletal muscle cell proliferation cell differentiation negative regulation of tumor necrosis factor production positive regulation of insulin secretion involved in cellular response to glucose stimulus response to ethanol positive regulation of gluconeogenesis positive regulation of vasodilation rhythmic process negative regulation of growth hormone secretion negative regulation of cell death negative regulation of reactive oxygen species biosynthetic process negative regulation of fibroblast apoptotic process regulation of energy homeostasis positive regulation of thyroid-stimulating hormone secretion
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0