Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • SI-CLP Antibodies

NeoBiotechnologies

SI-CLP/Chitinase Domain Containing 1 (CHID1) Monoclonal Antibody (CHID1/6368)

View all (6) SI-CLP antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite SI-CLP/Chitinase Domain Containing 1 (CHID1) Monoclonal Antibody (CHID1/6368)

  • Antibody Testing Data (2)
SI-CLP/Chitinase Domain Containing 1 (CHID1) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
SI-CLP/Chitinase Domain Containing 1 (CHID1) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

SI-CLP/Chitinase Domain Containing 1 (CHID1) Antibody (66005-MSM1-P1) in IHC (P)

Formalin-fixed, paraffin-embedded human prostate stained with SI-CLP/CHID1 Mouse Monoclonal Antibody (CHID1/6368) HIER: Tris/EDTA, pH9.0, 45 min. 2°C: HRP-polymer, 30 min. DAB, 5 min. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

SI-CLP/Chitinase Domain Containing 1 (CHID1) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
SI-CLP/Chitinase Domain Containing 1 (CHID1) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
SI-CLP/Chitinase Domain Containing 1 (CHID1) Monoclonal Antibody (CHID1/6368)

Product Details

66005-MSM1-P1

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

1-2 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1, kappa

Class

Monoclonal

Type

Antibody

Clone

CHID1/6368

Immunogen

Recombinant full-length human CHID1 protein
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

200 µg/mL

Purification

Protein A/G

Storage buffer

PBS with 0.05% BSA

Contains

0.05% sodium azide

Storage conditions

4°C

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Positive Control: colon HeLa cells. Human fetal gut liver or heart.

Cellular Location: Secreted. Lysosome.

Target Information

SI-CLP detects human Stabilin-interacting chitinase like protein (SI-CLP), a novel member of the Glyco-18-domain containing protein family. SI-CLP is a secretory protein which is up-regulated in response to interleukin-4 and dexamethasone in activated macrophages. SI-CLP is also expressed by T- and B-lymphocytes, and by cells of monocytic and epithelial origin. SI-CLP is the ligand for stabilin-1, an intracellular sorting receptor which sorts SI-CLP into late endosomes and secretory lysosomes in activated macrophages.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Chitinase domain-containing protein 1; FLJ42707; MGC3234; SI-CLP; stabilin-1 interacting chitinase-like protein; Stabilin-1-interacting chitinase-like protein

View more View less

Gene Aliases: CHID1; GL008; PSEC0104; SB139; SI-CLP; SICLP

View more View less

UniProt ID: (Human) Q9BWS9

View more View less

Entrez Gene ID: (Human) 66005

View more View less

Function(s)
chitinase activity protein binding chitin binding oligosaccharide binding
Process(es)
carbohydrate metabolic process chitin catabolic process innate immune response negative regulation of cytokine production involved in inflammatory response
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0