Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • RbAp46 Antibodies

Proteintech

RbAp46 Polyclonal Antibody

7 References
View all (25) RbAp46 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite RbAp46 Polyclonal Antibody

  • Antibody Testing Data (7)
RbAp46 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
RbAp46 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 7

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

RbAp46 Antibody (20365-1-AP) in ICC/IF

Immunofluorescent analysis of ( 10% Formaldehyde ) fixed A431 cells using 20365-1-AP (RbAp46 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

RbAp46 Antibody in Immunocytochemistry (ICC/IF)
RbAp46 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RbAp46 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RbAp46 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RbAp46 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RbAp46 Antibody in Western Blot (WB)
RbAp46 Antibody in Immunoprecipitation (IP)

Product Details

20365-1-AP

Applications
Tested Dilution
Publications

Western Blot (WB)

1:200-1:1,000
View 5 publications 5 publications

Immunohistochemistry (IHC)

-
View 2 publications 2 publications

Immunohistochemistry (Paraffin) (IHC (P))

1:20-1:200
-

Immunocytochemistry (ICC/IF)

1:50-1:500
-

Immunoprecipitation (IP)

0.5-4.0 µg
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Published species

Human

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Peptide

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.26 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice (domestic); Dry ice (international)

Product Specific Information

The antibody is specific to RBBP7.

Target Information

This protein is a ubiquitously expressed nuclear protein and belongs to a highly conserved subfamily of WD-repeat proteins. It is found among several proteins that binds directly to retinoblastoma protein, which regulates cell proliferation. The encoded protein is found in many histone deacetylase complexes, including mSin3 co-repressor complex. It is also present in protein complexes involved in chromatin assembly. This protein can interact with BRCA1 tumor-suppressor gene and may have a role in the regulation of cell proliferation and differentiation.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: G1/S transition control protein-binding protein; G1/S transition control protein-binding protein RbAp46; Histone acetyltransferase type B subunit 2; Histone-binding protein RBBP7; Nucleosome-remodeling factor subunit RBAP46; RBBP-7; retinoblastoma binding protein 7; Retinoblastoma-binding protein 7; Retinoblastoma-binding protein p46; retinoblastoma-binding protein RbAp46

View more View less

Gene Aliases: AA409861; AI173248; AU019541; BB114024; mRbAp46; RBAP46; RBBP7

View more View less

UniProt ID: (Human) Q16576, (Mouse) Q60973, (Rat) Q71UF4

View more View less

Entrez Gene ID: (Human) 5931, (Mouse) 245688, (Rat) 83712

View more View less

Function(s)
RNA binding histone deacetylase activity protein binding molecular_function
Process(es)
negative regulation of transcription from RNA polymerase II promoter DNA replication transcription, DNA-templated multicellular organism development cell proliferation histone deacetylation negative regulation of cell growth CENP-A containing nucleosome assembly negative regulation of gene expression, epigenetic response to steroid hormone cellular heat acclimation regulation of signal transduction by p53 class mediator chromatin remodeling regulation of transcription, DNA-templated chromatin modification negative regulation of transcription, DNA-templated
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0