Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Raptor Antibodies

Invitrogen

Raptor Recombinant Rabbit Monoclonal Antibody (9N0A4)

View all (27) Raptor antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Raptor Recombinant Rabbit Monoclonal Antibody (9N0A4)

  • Antibody Testing Data (3)
Raptor Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
Raptor Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Raptor Antibody (MA5-35742) in ICC/IF

Immunofluorescence analysis of Raptor in NIH-3T3 cells. Samples were incubated with Raptor Monoclonal antibody (Product # MA5-35742) using a dilution of 1:100 (40x lens). Blue: DAPI for nuclear staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Raptor Antibody in Immunocytochemistry (ICC/IF)
Raptor Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Raptor Antibody in Western Blot (WB)
Raptor Recombinant Rabbit Monoclonal Antibody (9N0A4)

Product Details

MA5-35742

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:2,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:100-1:1,000
-

Immunocytochemistry (ICC/IF)

1:50-1:200
-

ELISA (ELISA)

1 µg/mL
-
Product Specifications

Species Reactivity

Human, Mouse

Host/Isotype

Rabbit / IgG

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

9N0A4

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1236-1335 of human Raptor (Q8N122)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Affinity Chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol, 0.05% BSA

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2849642

Product Specific Information

Immunogen sequence: VRIFDPRMPE SVNVLQIVKG LTALDIHPQA DLIACGSVNQ FTAIYNSSGE LINNIKYYDG FMGQRVGAIS CLAFHPHWPH LAVGSNDYYI SVYSVEKRVR

Target Information

Regulatory associated protein of mTOR (RAPTOR) is a binding partner of target of rapamycin (TOR). It binds to 4EBP1 and p70alpha. The binding of raptor to mTOR is essential for the mTOR-catalyzed phosphorylation of 4EBP1 in vitro. It enhances the mTOR kinase activity toward p70alpha. It is also an essential scaffold for the mTOR-catalyzed phosphorylation of 4EBP1 and mediates TOR action in vivo.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: p150 target of rapamycin (TOR)-scaffold protein; p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats; Raptor; regulatory associated protein of MTOR, complex 1; Regulatory-associated protein of mTOR

View more View less

Gene Aliases: 4932417H02Rik; KIAA1303; KOG1; Mip1; mKIAA1303; Rap; RAPTOR; RPTOR

View more View less

UniProt ID: (Human) Q8N122

View more View less

Entrez Gene ID: (Human) 57521, (Mouse) 74370

View more View less

Function(s)
RNA polymerase III type 1 promoter DNA binding RNA polymerase III type 2 promoter DNA binding RNA polymerase III type 3 promoter DNA binding TFIIIC-class transcription factor binding protein binding protein kinase binding protein kinase activator activity protein binding, bridging protein complex binding 14-3-3 protein binding
Process(es)
positive regulation of endothelial cell proliferation cell cycle arrest regulation of cell size cell growth macroautophagy cellular response to nutrient levels TOR signaling positive regulation of TOR signaling activation of protein kinase activity positive regulation of transcription from RNA polymerase III promoter cellular response to amino acid stimulus positive regulation of protein serine/threonine kinase activity regulation of cellular response to heat
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0