Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • RXFP2 Antibodies

Invitrogen

RXFP2 Polyclonal Antibody

View all (9) RXFP2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite RXFP2 Polyclonal Antibody

  • Antibody Testing Data (4)
RXFP2 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
RXFP2 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

RXFP2 Antibody (PA5-98625) in ICC/IF

Immunofluorescent analysis of RXFP2 in HepG2 cells using a RXFP2 polyclonal antibody (Product # PA5-98625) at a dilution of 1:130. The cells were fixed in 4% formaldehyde, permeabilized using 0.2% Triton X-100 and blocked in 10% normal Goat Serum. The cells were then incubated with the antibody overnight at 4°C. The secondary antibody was Alexa Fluor 488-congugated Goat Anti-Rabbit IgG(H+L). Cells were counter-stained with DAPI. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

RXFP2 Antibody in Immunocytochemistry (ICC/IF)
RXFP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RXFP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RXFP2 Antibody in Western Blot (WB)
RXFP2 Polyclonal Antibody

Product Details

PA5-98625

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:5,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:200-1:500
-

Immunocytochemistry (ICC/IF)

1:50-1:200
-

ELISA (ELISA)

Assay-Dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Recombinant Human Relaxin receptor 2 protein (33-158AA)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

2.905 mg/mL

Purification

Protein G

Storage buffer

PBS, pH 7.4, with 50% glycerol

Contains

0.03% ProClin 300

Storage conditions

-20°C or -80°C if preferred

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2813238

Target Information

The activity of this relaxin receptor is mediated by G proteins leading to stimulation of adenylate cyclase and an increase of cAMP and may also be a receptor for Leydig insulin-like peptide. Expressed in embryonic and adult gonads of males and females, as well in male gubernarculum and in brain. Not detected in kidney, spleen and heart.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: G protein coupled receptor affecting testicular descent; G-protein coupled receptor 106; G-protein coupled receptor affecting testicular descent; leucine-rich repeat-containing G protein-coupled receptor 8; Leucine-rich repeat-containing G-protein coupled receptor 8; Relaxin family peptide receptor 2; Relaxin receptor 2; relaxin/insulin-like family peptide receptor 2

View more View less

Gene Aliases: GPR106; GREAT; INSL3R; LGR8; LGR8.1; RXFP2; RXFPR2

View more View less

UniProt ID: (Human) Q8WXD0

View more View less

Entrez Gene ID: (Human) 122042

View more View less

Function(s)
G-protein coupled receptor activity protein-hormone receptor activity peptide hormone binding
Process(es)
oocyte maturation adenylate cyclase-modulating G-protein coupled receptor signaling pathway adenylate cyclase-activating G-protein coupled receptor signaling pathway adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway negative regulation of cell proliferation male gonad development positive regulation of cAMP biosynthetic process negative regulation of apoptotic process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0