Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • RRP9 Antibodies

Abnova

RNU3IP2 Monoclonal Antibody (1E8)

View all (8) RRP9 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite RNU3IP2 Monoclonal Antibody (1E8)

  • Antibody Testing Data (1)
RNU3IP2 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
RNU3IP2 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

RNU3IP2 Antibody (H00009136-M01) in WB

RNU3IP2 monoclonal antibody (M01), clone 1E8. Western Blot analysis of RNU3IP2 expression in Jurkat. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

RNU3IP2 Antibody in Western Blot (WB)
RNU3IP2 Monoclonal Antibody (1E8)

Product Details

H00009136-M01

Applications
Tested Dilution
Publications

Western Blot (WB)

1-5 µg/mL
-

Immunohistochemistry (Paraffin) (IHC (P))

3 µg/mL
-

Immunocytochemistry (ICC/IF)

10-100 µg/mL
-

ELISA (ELISA)

10 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2b, kappa

Class

Monoclonal

Type

Antibody

Clone

1E8

Immunogen

RNU3IP2 (AAH01113, 1 a.a. approximately 475 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.5-1 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Product may be used with Western Blot (Cell lysate).

Immunogen sequence: MSATAAARKR GKPASGAGAG AGAGKRRRKA DSAGDRGKSK GGGKMNEEIS SDSESESLAP RKPEEEEEEE LEETAQEKKL RLAKLYLEQL RQQEEEKAEA RAFEEDQVAG RLKEDVLEQR GRLQKLVAKE IQAPASADIR VLRGHQLSIT CLVVTPDDSA IFSAAKDCSI IKWSVESGRK LHVIPRAKKG AEGKPPGHSS HVLCMAISSD GKYLASGDRS KLILIWEAQS CQHLYTFTGH RDAVSGLAFR RGTHQLYSTS HDRSVKVWNV AENSYVETLF GHQDAVAALD ALSRECCVTA GGRDGTVRVW KIPEESQLVF YGHQGSIDCI HLINEEHMVS GADDGSVALW GLSKKRPLAL QREAHGLRGE PGLEQPFWIS SVAALLNTDL VATGSHSSCV RLWQCGEGFR QLDLLCDIPL VGFINSLKFS SSGDFLVAGV GQEHRLGRWW RIKEARNSVC IIPLRRVPVP PAAGS

Target Information

The Ku autoantigen is a heterodimer of 70kDa (p70) and approximately 80kDa (p80) proteins. The p70/p80 dimer is important for function of a 460kDa DNA-dependent protein kinase that phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. Ku protein plays a role in cell signaling, proliferation, DNA repair, replication, transcriptional activation, and apoptosis.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: RNA, U3 small nucleolar interacting protein 2; RRP9 homolog; RRP9, small subunit (SSU) processome component, homolog; U3 small nucleolar ribonucleoprotein-associated 55 kDa protein; U3 small nucleolar RNA-interacting protein 2; U3 snoRNP-associated 55 kDa protein; U3 snoRNP-associated 55-kDa protein

View more View less

Gene Aliases: RNU3IP2; RRP9; U3-55K; U355K

View more View less

UniProt ID: (Human) O43818

View more View less

Entrez Gene ID: (Human) 9136

View more View less

Function(s)
U3 snoRNA binding poly(A) RNA binding
Process(es)
rRNA processing
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0