Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • RGS14 Antibodies

Proteintech

RGS14 Polyclonal Antibody

11 References
View all (21) RGS14 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite RGS14 Polyclonal Antibody

  • Antibody Testing Data (11)
RGS14 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
RGS14 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 11

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

RGS14 Antibody (16258-1-AP) in IHC (P)

Immunohistochemical analysis of paraffin-embedded mouse brain tissue slide using (Product # 16258-1-AP) (RGS14 antibody) at dilution of 1:5,000 (under 10x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

RGS14 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RGS14 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RGS14 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RGS14 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RGS14 Antibody in Western Blot (WB)
RGS14 Antibody in Western Blot (WB)
RGS14 Antibody in Western Blot (WB)
RGS14 Antibody in Western Blot (WB)
RGS14 Antibody in Western Blot (WB)
RGS14 Antibody in Western Blot (WB)
RGS14 Antibody in Immunoprecipitation (IP)
RGS14 Polyclonal Antibody

Product Details

16258-1-AP

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:2,000
View 4 publications 4 publications

Immunohistochemistry (IHC)

-
View 2 publications 2 publications

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:500
-

Immunocytochemistry (ICC/IF)

-
View 4 publications 4 publications

Immunoprecipitation (IP)

0.5-4.0 µg
View 1 publication 1 publication
Product Specifications

Species Reactivity

Human, Mouse

Published species

Human, Monkey, Mouse, Non-human primate, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

RGS14 Fusion Protein Ag9292 (217-566 aa encoded by BC014094)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.33 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice (domestic); Dry ice (international)

Product Specific Information

Immunogen sequence: KSLPLGVEE LGQLPPVEGP GGRPLRKSFR RELGGTANAA LRRESQGSLN SSASLDLGFL AFVSSKSESH RKSLGSTEGE SESRPGKYCC VYLPDGTASL ALARPGLTIR DMLAGICEKR GLSLPDIKVY LVGNEQALVL DQDCTVLADQ EVRLENRITF ELELTALERV VRISAKPTKR LQEALQPILE KHGLSPLEVV LHRPGEKQPL DLGKLVSSVA AQRLVLDTLP GVKISKARDK SPCRSQGCPP RTQDKATHPP PASPSSLVKV PSSATGKRQT CDIEGLVELL NRVQSSGAHD QRGLLRKEDL VLPEFLQLPA QGPSSEETPP QTKSAAQPIG GSLNSTTDSA L (217-566 aa encoded by BC014094)

Target Information

RGS14 is a member of the regulator of G-protein signaling family which contains one RGS domain, two Raf-like Ras-binding domains (RBDs), and one GoLoco domain. RGS14 attenuates the signaling activity of G-proteins by binding, through its GoLoco domain, to specific types of activated, GTP-bound G alpha subunits. Acting as a GTPase activating protein (GAP), the protein increases the rate of conversion of the GTP to GDP. The resulting hydrolysis allows the G alpha subunits to bind G beta/gamma subunit heterodimers, forming inactive G-protein heterotrimers, thereby terminating the signal. Alternate transcriptional splice variants of RGS14 have been observed but have not been thoroughly characterized. Further, RGS14 acts as a positive modulator of microtubule polymerisation and spindle organization through a G(i)-alpha-dependent mechanism; plays a role in cell division; is required for the nerve growth factor (NGF)-mediated neurite outgrowth, and is involved in stress resistance. RGS14 protein may also be involved in visual memory processing capacity and hippocampal-based learning and memory.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: rap1/rap2 interacting protein; RAP1/RAP2-interacting protein; Regulator of G-protein signaling 14; regulator of G-protein signalling 14; RGS14; RGSE; RPIP1

View more View less

Gene Aliases: RGS14; RPIP1

View more View less

UniProt ID: (Human) O43566, (Mouse) P97492

View more View less

Entrez Gene ID: (Human) 10636, (Mouse) 51791

View more View less

Function(s)
G-protein alpha-subunit binding receptor signaling protein activity GDP-dissociation inhibitor activity GTPase activator activity protein binding microtubule binding protein kinase binding receptor signaling complex scaffold activity GTPase activating protein binding GTPase regulator activity
Process(es)
nucleocytoplasmic transport response to oxidative stress cell cycle spindle organization chromosome segregation mitotic nuclear division signal transduction G-protein coupled receptor signaling pathway multicellular organism development learning long-term memory regulation of G-protein coupled receptor protein signaling pathway visual learning negative regulation of signal transduction zygote asymmetric cell division negative regulation of synaptic plasticity intracellular signal transduction termination of G-protein coupled receptor signaling pathway negative regulation of MAP kinase activity positive regulation of GTPase activity regulation of DNA-templated transcription in response to stress platelet-derived growth factor receptor signaling pathway positive regulation of neurogenesis cell division long-term synaptic potentiation negative regulation of ERK1 and ERK2 cascade
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0