Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • REG3A Antibodies

Invitrogen

REG3A Recombinant Rabbit Monoclonal Antibody (019)

View all (15) REG3A antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite REG3A Recombinant Rabbit Monoclonal Antibody (019)

  • Antibody Testing Data (2)
REG3A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
REG3A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

REG3A Antibody (MA5-29518) in IHC (P)

Immunohistochemical staining of human REG3A in human pancreatic carcinoma with REG3A Recombinant Rabbit Monoclonal Antibody (19) (Product # MA5-29518, 1:200, formalin-fixed paraffin embedded sections). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

REG3A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
REG3A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
REG3A Recombinant Rabbit Monoclonal Antibody (019)

Product Details

MA5-29518

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

1:100-1:500
-

ELISA (ELISA)

1:5,000-1:10,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

019

Immunogen

Recombinant Human REG3A protein (Met1-Asp175)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

PBS

Contains

no preservative

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2785368

Product Specific Information

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

This antibody has specificity for Human REG3A.

Target Information

REG3A encodes a pancreatic secretory protein that may be involved in cell proliferation or differentiation. It has similarity to the C-type lectin superfamily. The enhanced expression of this protein is observed during pancreatic inflammation and liver carcinogenesis.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: hepatocarcinoma-intestine-pancreas; Hepatointestinal pancreatic protein; HIP; HIP/PAP; Human proislet peptide; pancreatic beta cell growth factor; Pancreatitis-associated protein 1; PAP homologous protein; proliferation-inducing protein 34; proliferation-inducing protein 42; REG 3a; REG 3alpha; Reg III-alpha; Reg IIIalpha; REG-3-alpha; REG-3alpha; REG3 alpha; REG3alpha; regeneratin; regenerating islet-derived 3 alpha; Regenerating islet-derived protein 3-alpha; Regenerating islet-derived protein III-alpha

View more View less

Gene Aliases: HIP; HIP/PAP; INGAP; PAP; PAP-H; PAP1; PBCGF; REG-III; REG3; REG3A

View more View less

UniProt ID: (Human) Q06141

View more View less

Entrez Gene ID: (Human) 5068

View more View less

Function(s)
protein binding carbohydrate binding
Process(es)
acute-phase response heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules multicellular organism development cell proliferation positive regulation of keratinocyte proliferation negative regulation of keratinocyte differentiation positive regulation of wound healing
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0