Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • RBM15 Antibodies

OriGene

RBM15 Polyclonal Antibody

View all (17) RBM15 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite RBM15 Polyclonal Antibody

  • Antibody Testing Data (1)
RBM15 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
RBM15 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

RBM15 Antibody (TA590462) in WB

Western Blot: RBM15 Antibody - Samples: Lane 1: Marker: 250, 130, 95, 72, 55, 36, 28, 17, 11 / Lane 2: RT-4 / Lane 3: U-251MG sp / Lane 4: Human Plasma / Lane 5: Liver / Lane 6: Tonsil; Target weight: 107, 106, 3 (splice variants). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

RBM15 Antibody in Western Blot (WB)

Product Details

TA590462

Applications
Tested Dilution
Publications

Western Blot (WB)

1:5,000-1:20,000
-

Immunohistochemistry (IHC)

Assay-dependent
-

ELISA (ELISA)

1:100-1:2,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

DNA immunization. This antibody is specific for the Middle Region of the target protein.

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1.1 mg/mL

Purification

Protein A/G

Storage buffer

PBS

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

May function as an mRNA export factor, stimulating export and expression of RTE-containing mRNAs which are present in many retrotransposons that require to be exported prior to splicing. High affinity binding of pre-mRNA to RBM15 may allow targeting of the mRNP to the export helicase DBP5 in a manner that is independent of splicing-mediated NXF1 deposition, resulting in export prior to splicing. May be implicated in HOX gene regulation.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: one twenty two protein; one twenty-two; One-twenty two protein 1; RNA-binding motif protein 15; RNA-binding protein 15

View more View less

Gene Aliases: OTT; OTT1; RBM15; SPEN

View more View less

UniProt ID: (Human) Q96T37

View more View less

Entrez Gene ID: (Human) 64783

View more View less

Function(s)
nucleotide binding nucleic acid binding protein binding poly(A) RNA binding
Process(es)
mRNA splicing, via spliceosome patterning of blood vessels positive regulation of transcription of Notch receptor target viral process negative regulation of myeloid cell differentiation negative regulation of transcription, DNA-templated spleen development ventricular septum morphogenesis placenta blood vessel development
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0