Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Procollagen I C-Peptide Antibodies

QED Bioscience

Procollagen Type I C-Peptide (PIP) Monoclonal Antibody (PC8-7)

View all (5) Procollagen I C-Peptide antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Procollagen Type I C-Peptide (PIP) Monoclonal Antibody (PC8-7)

Product Details

42043-100UG

Applications
Tested Dilution
Publications

Western Blot (WB)

10 µg/mL
-

Immunohistochemistry (Paraffin) (IHC (P))

10 µg/mL
-

ELISA (ELISA)

0.2 µg/mL
-
Product Specifications

Species Reactivity

Bovine, Dog, Horse, Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

PC8-7

Immunogen

Human Procollagen Type I C-Peptide

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

0.5-1.0 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.4, with 1% BSA

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Reconstitute in 50 µL of distilled water (final concentration 2 mg/mL).

Target Information

Collagen type I is present in soft connective tissue and bone where it represents approximately 90% of the organix matrix. Collagen type I is produced from procollagen when two teloterminal propeptides are digested from the procursor and released into the circulation. The propeptide levels can be measured in serum as indicators of collagen production. Elevated values of these propeptides in serum are found in patients with various bone metabolism defects. Procollagen type I C-terminal propeptide (PICP) has a molecular weight of approximately 100 kDa and the normal range in serum depends on age and sex.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Alpha-1 type I collagen; alpha1(I) procollagen; C-terminal Collagen; collagen alpha 1 chain type I; Collagen alpha-1(I) chain; collagen alpha-1(I) chain preproprotein; collagen of skin, tendon and bone, alpha-1 chain; collagen, type I, alpha 1; PICP; pro-alpha-1 collagen type 1; Procol; Procollagen; type I collagen pre-pro-alpha1(I) chain; type I proalpha 1; type I procollagen alpha 1 chain

View more View less

Gene Aliases: BOS_18945; COL1A1; EDSC; OI1; OI2; OI3; OI4

View more View less

UniProt ID: (Dog) Q9XSJ7, (Human) P02452, (Bovine) P02453

View more View less

Entrez Gene ID: (Dog) 403651, (Horse) 100033877, (Human) 1277, (Bovine) 282187

View more View less

Function(s)
extracellular matrix structural constituent metal ion binding protein binding identical protein binding platelet-derived growth factor binding
Process(es)
skeletal system development blood vessel development osteoblast differentiation intramembranous ossification endochondral ossification blood coagulation visual perception sensory perception of sound positive regulation of epithelial to mesenchymal transition negative regulation of cell-substrate adhesion protein transport platelet activation extracellular matrix organization collagen fibril organization positive regulation of cell migration collagen catabolic process response to corticosteroid response to estradiol collagen biosynthetic process protein localization to nucleus tooth mineralization response to drug response to hydrogen peroxide response to peptide hormone skin morphogenesis cellular response to fibroblast growth factor stimulus tooth eruption positive regulation of transcription, DNA-templated embryonic skeletal system development regulation of immune response leukocyte migration response to cAMP response to hyperoxia face morphogenesis bone trabecula formation cartilage development involved in endochondral bone morphogenesis protein heterotrimerization cellular response to amino acid stimulus cellular response to mechanical stimulus cellular response to retinoic acid cellular response to vitamin E cellular response to tumor necrosis factor cellular response to epidermal growth factor stimulus cellular response to transforming growth factor beta stimulus positive regulation of canonical Wnt signaling pathway cellular response to fluoride
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0