Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • POLI Antibodies

Bethyl Laboratories

Pol Iota Polyclonal Antibody

View all (36) POLI antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Pol Iota Polyclonal Antibody

  • Antibody Testing Data (1)
Pol Iota Antibody in Immunoprecipitation (IP)
Group 53 Created with Sketch.
Pol Iota Antibody in Immunoprecipitation (IP)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Pol Iota Antibody (A301-303A) in IP

Detection of human Pol Iota by western blot of immunoprecipitates. Samples: Whole cell lysate (1 mg for IP, 20% of IP loaded) from HeLa cells. Antibodies: Affinity purified rabbit anti-Pol Iota antibody (Product # A301-303A) used for IP at 3 µg/mg lysate. Pol Iota was also immunoprecipitated by rabbit anti-Pol Iota antibody A301-304A, which recognizes a downstream epitope. For blotting immunoprecipitated Pol Iota, A301-304A was used at 1 µg/mL. Detection: Chemiluminescence with an exposure time of 30 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Pol Iota Antibody in Immunoprecipitation (IP)
Pol Iota Polyclonal Antibody

Product Details

A301-303A

Applications
Tested Dilution
Publications

Western Blot (WB)

Assay-dependent
-

Immunoprecipitation (IP)

2-5 µg/mg lysate
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Region between residue 640 and 690 of Human DNA polymerase iota.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

tris citrate/phosphate, pH 7-8

Contains

0.09% sodium azide

Storage conditions

4°C

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

The recommended shelf life for this product is 1 year from date of receipt.

Target Information

Error-prone DNA polymerase specifically involved in DNA repair. Plays an important role in translesion synthesis, where the normal high-fidelity DNA polymerases cannot proceed and DNA synthesis stalls. Favors Hoogsteen base-pairing in the active site. Inserts the correct base with high-fidelity opposite an adenosine template. Exhibits low fidelity and efficiency opposite a thymidine template, where it will preferentially insert guanosine. May play a role in hypermutation of immunoglobulin genes. Forms a Schiff base with 5'-deoxyribose phosphate at abasic sites, but may not have lyase activity.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: DNA polymerase iota; Eta2; polymerase (DNA directed) iota; polymerase (DNA) iota; polymerase (DNA-directed), iota; RAD 30B; RAD30; RAD30 homolog B

View more View less

Gene Aliases: POLI; RAD30B; RAD3OB

View more View less

UniProt ID: (Human) Q9UNA4

View more View less

Entrez Gene ID: (Human) 11201

View more View less

Function(s)
damaged DNA binding DNA-directed DNA polymerase activity protein binding metal ion binding
Process(es)
DNA replication DNA repair translesion synthesis error-prone translesion synthesis
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0