Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • TrkA/TrkB/TrkC Antibodies

Bioss

Phospho-TrkA/B/C (Tyr496, Tyr516, Tyr516) Polyclonal Antibody

View all (12) TrkA/TrkB/TrkC antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Phospho-TrkA/B/C (Tyr496, Tyr516, Tyr516) Polyclonal Antibody

  • Antibody Testing Data (1)
Phospho-TrkA/B/C (Tyr496, Tyr516, Tyr516) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
Phospho-TrkA/B/C (Tyr496, Tyr516, Tyr516) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Phospho-TrkA/B/C (Tyr496, Tyr516, Tyr516) Antibody (BS-5525R) in IHC (P)

Paraformaldehyde-fixed, paraffin embedded mouse brain; Antigen retrieval by boiling in sodium citrate buffer (pH6.0) for 15min; Block endogenous peroxidase by 3% hydrogen peroxide for 20 minutes; Blocking buffer (normal goat serum) at 37°C for 30min; Antibody incubation with TrkA (Tyr496) + TrkB (Tyr516) + TrKC (Tyr516) Antibody, Unconjugated (bs-5525R) at 1:500 overnight at 4°C, followed by a conjugated se... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Phospho-TrkA/B/C (Tyr496, Tyr516, Tyr516) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Phospho-TrkA/B/C (Tyr496, Tyr516, Tyr516) Polyclonal Antibody

Product Details

BS-5525R

Applications
Tested Dilution
Publications

Western Blot (WB)

Assay-dependent
-

Immunohistochemistry (Paraffin) (IHC (P))

1:500
-

Immunohistochemistry (Frozen) (IHC (F))

1:100-1:500
-

Immunocytochemistry (ICC/IF)

1:50-1:200
-

ELISA (ELISA)

1:500-1:1,000
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

KLH conjugated synthetic phosphopeptide derived from rat NTRK2 around the phosphorylation site of Tyr516.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

0.01M TBS, pH 7.4, with 1% BSA, 50% glycerol

Contains

0.02% ProClin 300

Storage conditions

-20°C

Shipping conditions

Ambient (domestic); Wet ice (international)

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: BDNF-tropomyosine receptor kinase B; BDNF/NT-3 growth factors receptor; ETS related protein-neurotrophic receptor tyrosine kinase fusion protein; ETV6-NTRK3 fusion; gp140trk; GP145-TrkB; GP145-TrkB/GP95-TrkB; GP145-TrkC; High affinity nerve growth factor receptor; neural receptor protein-tyrosine kinase (trkB); neural receptor protein-tyrosine kinase (trkC); Neurotrophic tyrosine kinase receptor type 1; Neurotrophic tyrosine kinase receptor type 2; Neurotrophic tyrosine kinase receptor type 3; neurotrophic tyrosine kinase, receptor, type 1; neurotrophic tyrosine kinase, receptor, type 2; neurotrophic tyrosine kinase, receptor, type 3; neurotrophic tyrosine receptor kinase type 2; neurotrophin-3; NT-3 growth factor receptor; Oncogene TRK; p140-TrkA; Slow nerve growth factor receptor; trk; Trk-A; Trk-B; Trk-C; TRK1-transforming tyrosine kinase protein; TrkA neurotrophin receptor; trkA proto-oncogene receptor; TrkB tyrosine kinase; TrkC tyrosine kinase; Tropomyosin-related kinase A; Tropomyosin-related kinase B; Tyrosine kinase receptor; Tyrosine kinase receptor A; tyrosine kinase receptor B; tyrosine kinase receptor C

View more View less

Gene Aliases: AW125844; C80751; gp145(trkC); GP145-TrkB; GP145-TrkB/GP95-TrkB; GP145-TrkC; MTC; NTRK1; NTRK2; NTRK3; Ntrk3_tv3; p140-TrkA; RATTRKB1; Tkr; Tkrb; TRK; Trk-A; trk-B; TRK1; TRKA; TRKB; TRKB1; TRKC

View more View less

UniProt ID: (Human) P04629, (Human) Q16620, (Human) Q16288, (Mouse) Q3UFB7, (Rat) P35739, (Mouse) P15209, (Rat) Q63604, (Mouse) Q6VNS1, (Rat) Q03351

View more View less

Entrez Gene ID: (Human) 4914, (Human) 4915, (Human) 4916, (Mouse) 18211, (Rat) 59109, (Mouse) 18212, (Rat) 25054, (Mouse) 18213, (Rat) 29613

View more View less

Function(s)
transmembrane receptor protein tyrosine kinase activity GPI-linked ephrin receptor activity neurotrophin p75 receptor binding protein binding ATP binding nerve growth factor receptor activity kinase binding protein homodimerization activity neurotrophin binding nerve growth factor binding receptor tyrosine kinase binding brain-derived neurotrophic factor binding brain-derived neurotrophic factor-activated receptor activity p53 binding neurotrophin receptor activity nucleotide binding protein kinase activity protein tyrosine kinase activity kinase activity transferase activity transferase activity, transferring phosphorus-containing groups ephrin receptor binding
Process(es)
protein phosphorylation transmembrane receptor protein tyrosine kinase signaling pathway multicellular organism development nervous system development axon guidance learning or memory programmed cell death involved in cell development response to activity phosphorylation peptidyl-tyrosine phosphorylation sensory perception of pain cell differentiation B cell differentiation response to nutrient levels response to nicotine nerve growth factor signaling pathway mechanoreceptor differentiation response to drug positive regulation of programmed cell death negative regulation of neuron apoptotic process positive regulation of angiogenesis protein autophosphorylation neurotrophin TRK receptor signaling pathway sympathetic nervous system development detection of temperature stimulus involved in sensory perception of pain detection of mechanical stimulus involved in sensory perception of pain positive regulation of NF-kappaB transcription factor activity response to electrical stimulus positive regulation of synapse assembly positive regulation of synaptic transmission, glutamatergic Sertoli cell development innervation axonogenesis involved in innervation behavioral response to formalin induced pain cellular response to nicotine cellular response to growth factor stimulus negative regulation of neuron death cellular response to nerve growth factor stimulus vasculogenesis neuron migration neuromuscular junction development learning long-term memory feeding behavior positive regulation of cell proliferation positive regulation of gene expression positive regulation of neuron projection development glutamate secretion positive regulation of phosphatidylinositol 3-kinase signaling regulation of metabolic process central nervous system neuron development cerebral cortex development neuron differentiation brain-derived neurotrophic factor receptor signaling pathway positive regulation of peptidyl-serine phosphorylation calcium-mediated signaling using intracellular calcium source neurotrophin signaling pathway regulation of GTPase activity regulation of MAPK cascade positive regulation of MAPK cascade retinal rod cell development regulation of neurotransmitter secretion oligodendrocyte differentiation peripheral nervous system neuron development positive regulation of axonogenesis regulation of dendrite development regulation of protein kinase B signaling retina development in camera-type eye long-term synaptic potentiation cellular response to amino acid stimulus positive regulation of glucocorticoid receptor signaling pathway negative regulation of anoikis activation of MAPK activity negative regulation of protein phosphorylation positive regulation of cell migration activation of protein kinase B activity positive regulation of apoptotic process neuron fate specification positive regulation of axon extension involved in regeneration negative regulation of astrocyte differentiation positive regulation of positive chemotaxis negative regulation of cell death lens fiber cell differentiation activation of GTPase activity positive regulation of actin cytoskeleton reorganization positive regulation of protein phosphorylation inflammatory response aging circadian rhythm response to light stimulus response to auditory stimulus cellular response to tumor necrosis factor cellular response to brain-derived neurotrophic factor stimulus heart development modulation by virus of host transcription response to ethanol ephrin receptor signaling pathway response to axon injury response to corticosterone cellular response to retinoic acid cochlea development negative regulation of cell proliferation response to radiation olfactory nerve development positive regulation of Ras protein signal transduction response to hydrostatic pressure positive regulation of ERK1 and ERK2 cascade activation of MAPKK activity microtubule-based movement positive regulation of GTPase activity phosphatidylinositol-mediated signaling
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0