Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Tau Antibodies

Invitrogen

Phospho-Tau (Thr231) Recombinant Rabbit Monoclonal Antibody (1G9)

View all (187) Tau antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Phospho-Tau (Thr231) Recombinant Rabbit Monoclonal Antibody (1G9)

  • Antibody Testing Data (1)
Phospho-Tau (Thr231) Antibody in Western Blot (WB)
Group 53 Created with Sketch.
Phospho-Tau (Thr231) Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Phospho-Tau (Thr231) Antibody (MA5-49767) in WB

Western blot analysis of Phospho-Tau (Thr231) in HeLa whole cell lysate (treated with Calyculin A or not) using a dilution of 1.63 µg/mL. Samples were incubated with Phospho-Tau (Thr231) monoclonal antibody (Product # MA5-49767). Secondary: Goat polyclonal to rabbit IgG at 1:50,000 dilution. Predicted band size: 46 KDa. Observed band size: 46 KDa. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Phospho-Tau (Thr231) Antibody in Western Blot (WB)

Product Details

MA5-49767

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:5,000
-

ELISA (ELISA)

Assay-dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

1G9

Immunogen

A synthesized peptide derived from human Phospho-MAPT (T231)

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.4 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.4, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C or -80°C if preferred

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_3092864

Target Information

Tau is a neuronal microtubule-associated protein found predominantly on axons. The function of Tau is to promote tubulin polymerization and stabilize microtubules. The C-terminus binds axonal microtubules while the N- terminus binds neural plasma membrane components, suggesting that tau functions as a linker protein between both. Axonal polarity is predetermined by TAU/MAPT localization (in the neuronal cell) in the domain of the cell body defined by the centrosome. The short isoforms allow plasticity of the cytoskeleton while the longer isoforms may preferentially play a role in its stabilization. In its hyper-phosphorylated form, Tau is the major component of paired helical filaments (PHF), the building block of neurofibrillary lesions in Alzheimer's diseases (AD) brain. Hyper-phosphorylation impairs the microtubule binding function of Tau, resulting in the destabilization of microtubules in AD brains, ultimately leading to the degeneration of the affected neurons. Numerous serine/threonine kinases phosphorylate Tau, including GSK-3beta, protein kinase A (PKA), cyclin-dependent kinase 5 (cdk5) and casein kinase II. Hyper-phosphorylated Tau is found in neurofibrillary lesions in a range and other central nervous system disorders such as Pick's disease, frontotemporal dementia, cortico-basal degeneration and progressive supranuclear palsy.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: FLJ31424; FTDP17; G protein beta1/gamma2 subunit-interacting factor 1; map tau; MGC138549; Microtubule-associated protein tau; microtubule-associated protein tau, isoform 4; microtubules; Neurofibrillary tangle protein; neurofibrillary tangles; Neuronal Marker; Paired helical filament-tau; PHF-tau; PHFtau; protein phosphatase 1, regulatory subunit 103; Tau-4; Tau5

View more View less

Gene Aliases: DDPAC; FTDP-17; MAPT; MAPTL; MSTD; MTBT1; MTBT2; PPND; PPP1R103; TAU

View more View less

UniProt ID: (Human) P10636

View more View less

Entrez Gene ID: (Human) 4137

View more View less

Function(s)
structural constituent of cytoskeleton protein binding microtubule binding SH3 domain binding enzyme binding protein kinase binding protein complex binding apolipoprotein binding protein phosphatase 2A binding lipoprotein particle binding
Process(es)
microtubule cytoskeleton organization neuron migration brain development female pregnancy response to nutrient adult walking behavior axo-dendritic transport intrinsic apoptotic signaling pathway in response to oxidative stress response to organic substance regulation of autophagy regulation of microtubule polymerization positive regulation of microtubule polymerization negative regulation of intracellular transport positive regulation of axon extension mitochondrion transport along microtubule axon extension generation of neurons regulation of microtubule-based movement
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0