Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • RNA pol II CTD Antibodies

NeoBiotechnologies

Phospho-RNA pol II CTD (Ser5) Recombinant Rabbit Monoclonal Antibody (POLR2A/9089R)

View all (46) RNA pol II CTD antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Phospho-RNA pol II CTD (Ser5) Recombinant Rabbit Monoclonal Antibody (POLR2A/9089R)

  • Antibody Testing Data (2)
Phospho-RNA pol II CTD (Ser5) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
Phospho-RNA pol II CTD (Ser5) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Phospho-RNA pol II CTD (Ser5) Antibody (5430-RBM4-P1) in IHC (P)

Formalin-fixed, paraffin-embedded human breast cancer stained with RNA Poll II Recombinant Rabbit Monoclonal Antibody (POLR2A/9089R). HIER: Tris/EDTA, pH9.0, 45 min. 2°C: HRP-polymer, 30 min. DAB, 5 min. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Phospho-RNA pol II CTD (Ser5) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Phospho-RNA pol II CTD (Ser5) Antibody in Western Blot (WB)
Phospho-RNA pol II CTD (Ser5) Recombinant Rabbit Monoclonal Antibody (POLR2A/9089R)

Product Details

5430-RBM4-P1

Applications
Tested Dilution
Publications

Western Blot (WB)

2-4 µg/mL
-

Immunohistochemistry (Paraffin) (IHC (P))

1-2 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG, kappa

Expression System

CHO cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

POLR2A/9089R

Immunogen

Ten repeats of synthetic peptide YSPTSPS using chemically synthesized phospho-Ser5
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

200 µg/mL

Purification

Protein A

Storage buffer

PBS with 0.05% BSA

Contains

0.05% sodium azide

Storage conditions

4°C

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Positive Control: HAP1 HepG2 K562 NIH3T3 or T47D cells. Human testis. PC3

Cellular Location: Nucleus.

Target Information

RNA polymerase II is the enzyme responsible for the synthesis of mRNA and many non-coding RNAs. POLR2A encodes RNA polymerase II subunit B1 (RPB1), the largest subunit of RNA polymerase II. The C-terminal domain (CTD) of RPB1 contains tandem heptapeptide repeats with the consensus sequence YSPTSPS. Phosphorylation and other modifications within this sequence influence the recruitment of factors that regulate the transcription cycle. Recruitment of RNA processing factors to the elongation complex is coordinated by the phosphorylation of Serine-2 and Serine-5 residues within the CTD.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 3'-5' exoribonuclease; DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kd subunit; DNA-directed RNA polymerase II subunit A; DNA-directed RNA polymerase II subunit RPB1; DNA-directed RNA polymerase III largest subunit; EC 2.7.7.6; MGC75453; Pol II S2p; polymerase (RNA) II (DNA directed) polypeptide A, 220kDa; polymerase (RNA) II subunit A; POLYMERASE II; RNA; RNA POL2; RNA polymerase II C-terminal domain; RNA polymerase II carboxy-terminal domain; RNA polymerase II subunit B1; RNA-directed RNA polymerase II subunit RPB1; RP02; RPB220; RPO21; SUA8

View more View less

Gene Aliases: hRPB220; hsRPB1; POLR2; POLR2A; POLRA; RPB1; RPBh1; RpIILS; RPO2; RPOL2

View more View less

UniProt ID: (Human) P24928

View more View less

Entrez Gene ID: (Human) 5430

View more View less

Function(s)
RNA polymerase II activity DNA binding DNA-directed RNA polymerase activity RNA-directed RNA polymerase activity protein binding ubiquitin protein ligase binding poly(A) RNA binding metal ion binding
Process(es)
mRNA splicing, via spliceosome transcription, RNA-templated transcription-coupled nucleotide-excision repair DNA-templated transcription, termination regulation of transcription, DNA-templated transcription from RNA polymerase II promoter transcription initiation from RNA polymerase II promoter transcription elongation from RNA polymerase II promoter 7-methylguanosine mRNA capping fibroblast growth factor receptor signaling pathway gene expression gene silencing by RNA positive regulation of RNA splicing somatic stem cell population maintenance snRNA transcription from RNA polymerase II promoter positive regulation of viral transcription
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0