Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • HSF1 Antibodies

Invitrogen

Phospho-HSF1 (Ser326) Recombinant Rabbit Monoclonal Antibody (2A7)

View all (45) HSF1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Phospho-HSF1 (Ser326) Recombinant Rabbit Monoclonal Antibody (2A7)

  • Antibody Testing Data (1)
Phospho-HSF1 (Ser326) Antibody in Western Blot (WB)
Group 53 Created with Sketch.
Phospho-HSF1 (Ser326) Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Phospho-HSF1 (Ser326) Antibody (MA5-33167) in WB

Western Blot analysis of Phospho-HSF1 (Ser326) using a Phospho-HSF1 (Ser326) Monoclonal antibody (Product # MA5-33167) at a concentration of 0.8 µg/mL. Positive WB detected in: Hela whole cell lysate. All lanes: Phospho-HSF1 antibody at 0.8µg/ml. A secondary Goat polyclonal antibody to rabbit IgG was applied at a 1:50,000 dilution. Observed band size: 82 kDa. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Phospho-HSF1 (Ser326) Antibody in Western Blot (WB)
Phospho-HSF1 (Ser326) Recombinant Rabbit Monoclonal Antibody (2A7)

Product Details

MA5-33167

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:5,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:200
-

ELISA (ELISA)

Assay-Dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

2A7

Immunogen

A synthesized peptide derived from human Phospho-HSF1 (S326)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.55 mg/mL

Purification

Protein G, Antigen affinity chromatography

Storage buffer

PBS, pH 7.4, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C or -80°C if preferred

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2811983

Target Information

All organisms respond to elevated temperatures and a variety of environmental stresses by rapid synthesis of heat shock RNAs and proteins. The regulation of heat shock gene transcription is mediated by the transcriptional activator, heat shock factor (HSF), which binds to heat shock response elements (HSEs). These HSEs are found as three repeats of a 5-nucleotide {nGAAn} module, arranged in alternating orientation and present upstream of all heat shock genes. The HSEs are highly conserved among species yet HSF purified from yeast, Drosophila and human have different molecular weights and the proteins do not show significant immunological cross reaction. Two HSFs have been identified in human cells, HSF1 and HSF2, which bind to the same HSEs and have 38% sequence identity. These factors are activated by distinct stimuli, HSF1 is responsive to classical stress signals such as heat, heavy metals and oxidative reagents, whereas HSF2 is activated during hemin-mediated differentiation of human erythroleukemia cells. HSF1 exists constitutively in the cytoplasm and the nucleus of unstressed cells as a monomer which lacks DNA binding activity. Through an unknown signal generated during stress, HSF1 becomes activated to a nuclear localized, trimeric state which binds to DNA. The phosphorylation of HSF1 is necessary for maximal transcription of heat shock genes.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Heat shock factor protein 1; Heat shock transcription factor 1; HSF 1; HSTF 1

View more View less

Gene Aliases: HSF1; HSTF1

View more View less

UniProt ID: (Human) Q00613

View more View less

Entrez Gene ID: (Human) 3297

View more View less

Function(s)
RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II core promoter sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding RNA polymerase II intronic transcription regulatory region sequence-specific DNA binding chromatin binding transcription factor activity, sequence-specific DNA binding protein binding
Process(es)
negative regulation of transcription from RNA polymerase II promoter embryonic placenta development protein phosphorylation defense response female meiotic division spermatogenesis negative regulation of cell proliferation mRNA transcription response to lipopolysaccharide negative regulation of tumor necrosis factor production cellular response to heat positive regulation of multicellular organism growth positive regulation of transcription from RNA polymerase II promoter embryonic process involved in female pregnancy regulation of cellular response to heat
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0